Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Home  >  Heating Mantles
Heating Mantles
Heating Mantles
An electric laboratory heating mantle is used to uniformly heat various lab and glassware vessels. If you are looking for an accurate and easy-to-use heating solution this style of laboratory heating mantle is a good alternative to heating baths. This type of laboratory heating mantle is even great for those hard-to-fit flasks and funnels. You can choose from flexible fabric or canister-style—both types incorporate a flexible and insulated heating device which allows you to place your containers directly on the heating element without substantially increasing the risk of the glassware shattering. They do not have built-in temperature controllers and may require the use of a separate temperature or power controller. Your laboratory heating mantle is available with a wide variety of shape, size, wattage, and temperature control options. Selecting the right laboratory heating mantle for your needs
 Product Detail
Sort
 Availability
 Price
Glas-Col Flexible fabric heating mantle, 500 mL, 270 watts, 120 VAC
  • Product Type: Heating Mantle
  • Watts: 270
  • Flask size: 500 mL
EW-36225-30
In stock.
$229.00 USD/EACH
$194.65 USD/EACH
Qty:
Electrothermal Electromantle, 1000 mL capacity, 115 VAC
  • Product Type: Heating Mantle
  • Watts: 300
  • Flask size: 1000 mL
EW-36002-16
In stock.
$522.00 USD/EACH
$443.70 USD/EACH
Qty:
Glas-Col heating mantle, 600 mL, 325 watts, 115 VAC
  • Product Type: Heating Mantle
  • Watts: 325
  • Flask size: 600 mL
EW-36235-00
Usually ships in 14 days.
$289.00 USD/EACH
$245.65 USD/EACH
Qty:
Glas-Col Flexible fabric heating mantle, 5000 mL, 600 watts, 120 VAC
  • Product Type: Heating Mantle
  • Watts: 600
  • Flask size: 5000 mL
EW-36225-70
In stock.
$380.00 USD/EACH
$323.00 USD/EACH
Qty:
Glas-Col Flexible fabric heating mantle, 1000 mL, 380 watts, 120 VAC
  • Product Type: Heating Mantle
  • Watts: 380
  • Flask size: 1000 mL
EW-36225-40
In stock.
$263.00 USD/EACH
$223.55 USD/EACH
Qty:
Heating Mantle for 2000 mL beaker, 550 watts, 115 VAC
  • Product Type: Heating Mantle
  • Watts: 550
  • Flask size: 2000 mL
EW-36227-16
In stock.
$370.00 USD/EACH
$314.50 USD/EACH
Qty:
Glas-Col aluminum heating mantle, 1000 mL, 380 watts, 115 VAC
  • Product Type: Heating Mantle
  • Watts: 380
  • Flask size: 500 mL
EW-36233-20
In stock.
$317.00 USD/EACH
$269.45 USD/EACH
Qty:
Glas-Col aluminum heating mantle, 3000 mL, 500 watts, 115 VAC
  • Product Type: Heating Mantle
  • Watts: 500
  • Flask size: 3000 mL
EW-36233-40
Usually ships in 9 days.
$407.00 USD/EACH
$345.95 USD/EACH
Qty:
Glas-Col Flexible fabric heating mantle, 100 mL, 80 watts, 120 VAC
  • Product Type: Heating Mantle
  • Watts: 80
  • Flask size: 100 mL
EW-36225-02
In stock.
$185.00 USD/EACH
$157.25 USD/EACH
Qty:
Electrothermal Aluminum Heating Mantle, 250 mL Flask; 115 VAC
  • Product Type: Heating Mantle
  • Watts: 200
  • Flask size: 250 mL
EW-03012-31
In stock.
$377.00 USD/EACH
$320.45 USD/EACH
Qty:
Spherical Heating Mantle, 500 mL Flask, 0 W Upper/270 W Lower; 115 VAC
  • Product Type: Heating Mantle
  • Watts: 270
  • Flask size: 500 mL
EW-36226-10
Usually ships in 13 days.
$396.00 USD/EACH
$336.60 USD/EACH
Qty:
Electrothermal Aluminum Heating Mantle, 500 mL Flask; 115 VAC
  • Product Type: Heating Mantle
  • Watts: 300
  • Flask size: 500 mL
EW-03012-41
In stock.
$411.00 USD/EACH
$349.35 USD/EACH
Qty:
Glas-Col 100E O545 Support For Glas-Col Flexible Fabric Heating Mantle, 3000 mL
  • Product Type: Specialty Supports
  • Brand: Glas-Col
  • Manufacturer number: 100E O545
EW-36225-86
Usually ships in 12 days.
$123.00 USD/EACH
$104.55 USD/EACH
Qty:
Glas-Col 100E O550 Support For Glas-Col Flexible Fabric Heating Mantle, 5000 mL
  • Product Type: Specialty Supports
  • Brand: Glas-Col
  • Manufacturer number: 100E O550
EW-36225-87
Usually ships in 12 days.
$127.00 USD/EACH
$107.95 USD/EACH
Qty:
Glas-Col heating mantle, 1000 mL, 430 watts, 115 VAC
  • Product Type: Heating Mantle
  • Watts: 430
  • Flask size: 1000 mL
EW-36235-10
In stock.
$305.00 USD/EACH
$259.25 USD/EACH
Qty:
273 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#
#PRODUCTHEADER#
#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Heating Mantles