Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Safety Products
>
Radiation Detectors
> Heavy-Duty Surface Radiation Monitor
Heavy-Duty Surface Radiation Monitor
Detect alpha, beta, and gamma radiation in mR/hr and cpm
Heavy-duty construction ensures that this monitor will stand the rigors of daily use. Thin, recessed end window permits easy penetration of ions and is protected by a sturdy aluminum grill. The speaker and Geiger-Müller (GM) tube are housed in a rugged case. Replaceable neoprene feet on bottom of monitor let you place unit on bench or desktop—bottom face of instrument will not be contaminated.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Surface Radiation Monitor
Display analog bar graph,Model TBM-3S
EW-81920-00
In stock.
$813.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Heavy-Duty Surface Radiation Monitor
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Safety Products
>
Radiation Detectors
> Heavy-Duty Surface Radiation Monitor
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity