United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Display:
LED
(1)
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Heating Equipment
>
Sterilizers
> Heidolph Tabletop Autoclaves
Heidolph Tabletop Autoclaves
Offer fast, safe, dependable steam sterilization for your lab
Sterilize liquids, media, instruments, glassware, clothing, or waste
Models available with printer to meet GLP requirements
No plumbing or hard-wiring required
These compact benchtop autoclaves are extremely versatile, offering analog or electronic controls and providing a safe and convenient method for the sterilization of solids and liquids. Both analog and electronic models feature 316 stainless steel sterilization chambers and removable trays that resist corrosion. The continuous pressure purge helps eliminate cool air pockets within the chamber, ensuring thorough, even sterilization. A heatinsulated door with double-locking feature prevents opening while the chamber is under pressure to help minimize dangers to lab personnel. All models feature over-temperature cutoff, over-pressure valve, and a low-water level alarm which helps prevent damage to heating elements by signaling the need to refill the reservoir. A drying cycle helps prevent rusting of delicate instruments and helps dry clothing or absorbent materials.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital with Printer 0.8 cu ft, 120VAC
Product Type Sterilizer,Capacity cu ft 0.8,Chamber size " D 18
EW-10780-11
Call 1-888-358-4705 for availability.
$7,920.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Heidolph Tabletop Autoclaves
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Analog 0.8 cu ft, 220VAC
Product Type Sterilizer,Capacity cu ft 0.8,Chamber size " D 18
EW-10780-05
Call 1-888-358-4705 for availability.
$6,210.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Heidolph Tabletop Autoclaves
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Heating Equipment
>
Sterilizers
> Heidolph Tabletop Autoclaves
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title