United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Availability:
In Stock
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Centrifuges
>
Clinical Centrifuges
> Hermle Compact Variable-speed Universal Centrifuges
Hermle Compact Variable-speed Universal Centrifuges
Rotor choice offers flexibilty - process capacities from 0.2 to 50-mL
Set and view speed in both rpm and g-force
Adjust acceleration and deceleration rates to ten different levels
Imbalance sensor disables centrifuge and flashes an error message if unstable operating conditions occur
Control all settings with single ergonomic adjustment knob. Unit can be set for timed run or continuous operation. Preselect rotor code for rpm/RCF conversion. Splash proof LCD display lets you view preset and actual values. Accepts 6 5mL tubes, 18 1.5mL, 12 15mL or 6 50mL tubes depending on rotor assembly.
Durable ABS-plastic housing resists chemical spills and limits noise to under 60 dBA. Lid lock system improves safety by preventing the centrifuge from operating while the lid is open.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Compact Variable-Speed Universal Centrifuges; 115V; 50/60Hz
Product Type Centrifuge,Capacity Dependent upon rotor (see accessories),Max rpm 6000
EW-17306-01
In stock.
$1,850.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hermle Compact variable-speed universal centrifuge
Compact Variable-Speed Universal Centrifuges; 230V; 50/60Hz
Product Type Centrifuge,Capacity Dependent upon rotor,Max rpm 6000
EW-17306-06
Usually ships in 9 days.
$1,850.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hermle Compact Variable-speed Universal Centrifuges
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Centrifuges
>
Clinical Centrifuges
> Hermle Compact Variable-speed Universal Centrifuges
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title