Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Safety Products
>
Gloves
>
Disposable Gloves
>
Disposable Nitrile Gloves
> High Five A+ Aloe Nitrile Gloves
High Five A+ Aloe Nitrile Gloves
Formulated with skin-protecting aloe
Powder-free gloves feature a unique aloe coating on the interior glove surface for a comfortable, soothing feel against bare hands. Aloe prevents the drying and chapping that plagues frequent glove wearers.
Gloves are latex-free and contain no natural rubber proteins. Synthetic formulation offers good chemical and puncture resistance. Beaded cuff ensures easy donning. Ambidextrous.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
High Five A+ Aloe
Nitrile Gloves, Medium, 100/bx
Product Type Gloves,Material Nitrile,Color Green,Thickness mil 5
EW-86480-12
In stock.
$18.00 USD/BOX
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
High Five A+ Aloe Nitrile Gloves
High Five A+ Aloe
Nitrile Gloves, Large, 100/bx
Product Type Gloves,Material Nitrile,Thickness mil 5,Color Green
EW-86480-14
In stock.
$18.00 USD/BOX
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
High Five A+ Aloe Nitrile Gloves
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Safety Products
>
Gloves
>
Disposable Gloves
>
Disposable Nitrile Gloves
> High Five A+ Aloe Nitrile Gloves
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity