log in or register
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
High-Pressure Piston Pumps with Series Q Pump Heads
High-Pressure Piston Pumps with Series Q Pump Heads
  • Get high pressures, accurate dispensing, and great repeatability—all with one pump
  • Drives feature six dispensing modes—volume within a preset time, volume in steps, time, intermittent in time or volume, drip-free, or valve operation
  • Control speed, start/stop flow, or change fluid direction manually or remotely via an analog input signal or RS-232 interface
  • Series Q piston pump heads (included) have no valves to clog or wear out
Up to four sets of operating parameters can be stored in the pump (for pumps with Series Q pump heads only). RS-232 interface allows for control of pump via a PC or for cascading up to eight pumps. Housing is IP65 rated for Series Q pumps and IP30 rated for Series R pumps.
 Product Detail
Sort
 Availability
 Price
EW-78021-12 High-pressure piston pumps with Series Q pump head
Ismatec high-pressure pump with Series Q pump heads, 0.064 to 77 mL/min, 100 psi, ceramic piston/316 SS body/ceramic liner
Product Type Piston Metering Pumps,Max pressure 100 psi,Min flow rate mL/min 0.13
EW-78021-12
Call 1-888-358-4705 for availability.
$3,788.00 USD/EACH
Qty:
EW-78021-18 High-pressure piston pumps with Series Q pump head
Ismatec high-pressure pump with Series Q pump heads, 0.14 to 173 mL/min, 100 psi, 316 SS piston/316 SS body/ carbon liner
Product Type Piston Metering Pumps,Max pressure 100 psi,Min flow rate mL/min 0.29
EW-78021-18
Call 1-888-358-4705 for availability.
$3,811.00 USD/EACH
Qty:
EW-78021-20 High-pressure piston pumps with Series Q pump head
Ismatec high-pressure pump with Series Q pump heads, 0.14 to 173 mL/min, 60 psi, ceramic piston/Kynar body/ceramic liner
Product Type Piston Metering Pumps,Max pressure 60 psi,Min flow rate mL/min 0.29
EW-78021-20
Call 1-888-358-4705 for availability.
$3,667.00 USD/EACH
Qty:
EW-78021-10 High-pressure piston pumps with Series Q pump head
Ismatec high-pressure pump with Series Q pump heads, 0.064 to 77 mL/min, 60 psi, ceramic piston/Kynar body/carbon liner
Product Type Piston Metering Pumps,Max pressure 60 psi,Min flow rate mL/min 0.13
EW-78021-10
Call 1-888-358-4705 for availability.
$3,668.00 USD/EACH
Qty:
EW-78021-26 High-pressure piston pumps with Series Q pump head
Ismatec high-pressure pump with Series Q pump heads, 0.14 to 173 mL/min, 100 psi, ceramic piston/316 SS body/carbon liner
Product Type Piston Metering Pumps,Max pressure 100 psi,Min flow rate mL/min 0.29
EW-78021-26
Call 1-888-358-4705 for availability.
$3,811.00 USD/EACH
Qty:
EW-78021-28 High-pressure piston pumps with Series Q pump head
EW-78021-28
Call 1-888-358-4705 for availability.
$3,921.00 USD/EACH
Qty:
EW-78021-08 High-pressure piston pumps with Series Q pump head
Ismatec high-pressure pump with Series Q pump heads, 0.064 to 77 mL/min, 60 psi, ceramic piston/Kynar body/ceramic liner
Product Type Piston Metering Pumps,Max pressure 60 psi,Min flow rate mL/min 0.13
EW-78021-08
Call 1-888-358-4705 for availability.
$3,667.00 USD/EACH
Qty:
EW-78021-30 High-pressure piston pumps with Series Q pump head
EW-78021-30
Call 1-888-358-4705 for availability.
$3,921.00 USD/EACH
Qty:
EW-78021-16 High-pressure piston pumps with Series Q pump head
Ismatec high-pressure pump with Series Q pump heads, 0.14 to 173 mL/min, 60 psi, 316 SS piston/Kynar body/carbon liner
Product Type Piston Metering Pumps,Max pressure 60 psi,Min flow rate mL/min 0.29
EW-78021-16
Call 1-888-358-4705 for availability.
$3,688.00 USD/EACH
Qty:
EW-78021-00 High-pressure piston pumps with Series Q pump head
Ismatec high-pressure pump with Series Q pump heads, 0.016 to 19 mL/min, 100 psi, 316 SS piston/316 SS body/ carbon liner
Product Type Piston Metering Pumps,Max pressure 100 psi,Min flow rate mL/min 0.04
EW-78021-00
Call 1-888-358-4705 for availability.
$3,944.00 USD/EACH
Qty:
EW-78021-02 High-pressure piston pumps with Series Q pump head
Ismatec high-pressure pump with Series Q pump heads, 0.016 to 19 mL/min, 60 psi, 316 SS piston/Kynar body/carbon liner
Product Type Piston Metering Pumps,Max pressure 60 psi,Min flow rate mL/min 0.04
EW-78021-02
Call 1-888-358-4705 for availability.
$3,695.00 USD/EACH
Qty:
EW-78021-06 High-pressure piston pumps with Series Q pump head
Ismatec high-pressure pump with Series Q pump heads, 0.064 to 77 mL/min, 100 psi, 316 SS piston/316 SS body/ carbon liner
Product Type Piston Metering Pumps,Max pressure 100 psi,Min flow rate mL/min 0.13
EW-78021-06
Call 1-888-358-4705 for availability.
$3,811.00 USD/EACH
Qty:
EW-78021-24 High-pressure piston pumps with Series Q pump head
Ismatec high-pressure pump with Series Q pump heads, 0.14 to 173 mL/min, 100 psi, ceramic piston/316 SS body/ceramic liner
Product Type Piston Metering Pumps,Max pressure 100 psi,Min flow rate mL/min 0.29
EW-78021-24
Call 1-888-358-4705 for availability.
$3,788.00 USD/EACH
Qty:
EW-78021-22 High-pressure piston pumps with Series Q pump head
Ismatec high-pressure pump with Series Q pump heads, 0.14 to 173 mL/min, 60 psi, ceramic piston/Kynar body/carbon liner
Product Type Piston Metering Pumps,Max pressure 60 psi,Min flow rate mL/min 0.29
EW-78021-22
Call 1-888-358-4705 for availability.
$3,668.00 USD/EACH
Qty:
EW-78021-14 High-pressure piston pumps with Series Q pump head
Ismatec high-pressure pump with Series Q pump heads, 0.064 to 77 mL/min, 100 psi, ceramic piston/316 SS body/carbon liner
Product Type Piston Metering Pumps,Max pressure 100 psi,Min flow rate mL/min 0.13
EW-78021-14
Call 1-888-358-4705 for availability.
$3,811.00 USD/EACH
Qty:
16 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title