United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Tube OD (mm):
1 - 4
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Fittings
>
Chromatography fittings
>
PTFE Valves
>
Micro Tubing Distribution Valves
> High-pressure micro tubing valves
High-pressure micro tubing valves
Choose 4- or 5-port valves for use with 1 to 4 mm OD tubing (see description under “Micro tubing valves” above). Standard valves come with caps and VITON O-rings—withstand fluid pressures to 50 psi. The high-pressure valve withstands fluid pressures to 500 psi—order grippers separately on page 294.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
High-pressure valve, PTFE, 4 port, 90
adjacent ports connection, for 1/16" or 1/8" OD tubing
Product Type Chromatography fittings,Material PTFE,Brand Diba
EW-06471-76
Call 1-888-358-4705 for availability.
$270.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
High-pressure micro tubing valves
High-pressure micro tubing valves; 5 ports; one port to inlet; tube OD 1 to 4 mm
Product Type Chromatography fittings,Tube OD mm 1 - 4,Material PTFE
EW-06471-75
Call 1-888-358-4705 for availability.
$260.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
High-pressure micro tubing valves
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Fittings
>
Chromatography fittings
>
PTFE Valves
>
Micro Tubing Distribution Valves
> High-pressure micro tubing valves
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title