Honeywell Impact Multigas Monitors and Cartridges
Honeywell Impact Multigas Monitors and Cartridges
  • Replaceable sensor cartridges minimize maintenance downtime and cost
  • Increasingly rapid flashing, ultra-bright alarm LEDs
  • Three levels of instantaneous alarm
  • Time-weighted average alarms on toxic
The Impact multigas monitor is capable of monitoring up to four gases. Select 12-month disposable cartridges based on gases present. Check gas prior to entering a confined space using a drop line, hand aspirator, or optional pump. Patented Surecell sensors ensure stable, accurate, and reliable operation. A large, multi-lingual, backlit display with intuitive menu system simplifies use and minimizes operator training. The durable, rugged impact-resistant housing is waterproof to IP67 rating, yet small and lightweight for use in tight spaces. Meters feature a periodic “confidence” flash and beep, and powerful two-tone audible alarm.
Approval: UL and CSA Class I, Div I, Groups A, B, C, and D
Note: Cartridges are required and sold separately.
 Product Detail
Sort
 Availability
 Price
EW-81948-40 Honeywell Impact Multigas Monitor
Honeywell Impact Multigas Monitor
Product Type Multigas Monitors/Detectors,Brand Honeywell
EW-81948-40
Usually ships in 10 days.
$582.00 USD/EACH
Qty:
1 results (showing 1 - 1 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title