Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Life Science
>
Forensics
>
Safety & Response Products
>
Air Sampling and Gas Analysis
>
Gas Detection Instruments
>
Multigas Gas Detection
> Honeywell Impact Pro Multigas Monitors and Cartridges
Honeywell Impact Pro Multigas Monitors and Cartridges
Replaceable sensor cartridges minimize maintenance downtime and cost
Increasingly rapid flashing, ultra-bright alarm LEDs
Three levels of instantaneous alarm
Time-weighted average alarms on toxic
These multigas monitors are capable of monitoring up to four gases (multigas cartridges are sold separately). Check gas prior to entering a confined space using a drop line, hand aspirator, or built-in sampling pump. Patented Surecell
sensors ensure stable, accurate, and reliable operation. A large, multi-lingual, backlit display with intuitive menu system simplifies use and minimizes operator training. The durable, rugged impact-resistant housing is waterproof to IP67 rating, yet small and lightweight for use in tight spaces. Meters feature a periodic "confidence" flash and beep, and powerful two-tone audible alarm. Impact Pro features a built-in sampling pump, vibrating alarm, and multilingual menu. Link two Impact Pro detectors together using the optional Safelink cable to safely monitor personnel in a space.
Note:
Sensor cartridges are required and sold separately.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Honeywell Impact Pro Multigas Monitor
Product Type Multigas Monitors/Detectors,Brand Honeywell
EW-81948-41
Usually ships in 15 days.
$813.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Honeywell Impact Pro Multigas Monitors and Cartridges
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Life Science
>
Forensics
>
Safety & Response Products
>
Air Sampling and Gas Analysis
>
Gas Detection Instruments
>
Multigas Gas Detection
> Honeywell Impact Pro Multigas Monitors and Cartridges
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity