Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

Horiba Oil Content Analyzer

Horiba Oil Content Analyzer

  • Routine analysis of oil residuals
The Horiba OCMA-350 analyzer simplifies measurement of oil in water or soil for a wide variety of applications. Use this precision instrument for surveying water quality and hazardous waste sites, industrial wastewater monitoring, post-cleaning discharge analysis, and measuring adherent oil residuals on textiles and metal parts.

Use S-316 solvent to quantitatively extract sample. The oil / solvent solution is then simply transferred to the OCMA-350 analyzer for single-button measurement. Analysis is based on infrared (IR) spectral data, with peak integration (at approximately 3.4 µm) to determine oil concentration. S-316 solvent is non-absorbant at this IR wavelength, so only oil is measured. Accurately measures oil down to 0.1 ppm. Auto-calibrates after initial standard reading.

Use built-in time stamping to document results. Read results on LCD or transfer data using parallel printer port or to computer via RS-232. S-316 solvent can be recycled using Solvent Reclaimer SR-300 (82900-20), sold separately below.

What’s included: Quartz cell with cap, power cord, B-heavy oil (standard), 2 amp fuse, 10 mL syringe and 25 µL syringe.
 Product Detail
Sort
 Availability
 Price
EW-82900-00 Horiba Oil Content Analyzer, 115 V

Horiba Oil Content Analyzer, 115 V

Product Type Petroleum Testing Spectroscopes,Output RS-232C and centronics printer port,Display LCD
EW-82900-00
Usually ships in 7 days.
$7,000.00 USD/EACH

Qty:

Representative photo only Horiba Oil Content Analyzer 230 V

Horiba Oil Content Analyzer, 230 V

Product Type Petroleum Testing Spectroscopes,Output RS-232C and centronics printer port,Display LCD
EW-82900-05
Usually ships in 12 days.
$7,000.00 USD/EACH

Qty:

2 results (showing 1 - 2 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title