log in or register
Price Range:
Availability:
Material:
Acetal (4)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Fittings > Quick-Disconnect Fittings > Miniature Quick-Disconnect Fittings > Hose Barbs (female fitting halves)
Hose Barbs (female fitting halves)
Hose Barbs (female fitting halves)
Valveless fittings are completely interchangeable—for a complete fitting, order fitting bodies and inserts (female and male quick-disconnect halves) separately. Fittings halves seal with a twist; replaceable O-rings eliminate leaks. A protective shroud on 1/16" fittings prevents barb breakage.
Choose from DELRIN acetal or polypropylene (PP). DELRIN fittings have Buna N seals and an operating temp range of –40 to 180°F (–40 to 82°C). PP fittings have EPR seals and an operating temp range of 32 to 230°F (0 to 110°C). Both types are rated to 100 psi for vacuum or pressure applications. Pack of 10*.
 Product Detail
Sort
 Availability
 Price
Representative photo only Miniature Quick Disconnect Hose Barb Female Valved 1 16 ID
Miniature Quick-Disconnect Hose Barb (Female); Valved, 1/16" ID
Product Type Miniature Quick-Disconnects,Tubing ID in. 0.0625,Material Acetal
EW-06363-55
In stock.
$110.00 USD/PACKAGE
Qty:
Representative photo only Miniature Quick Disconnect Hose Barb Female Valved 1 8 ID
Miniature Quick-Disconnect Hose Barb (Female); Valved, 1/8" ID
Product Type Miniature Quick-Disconnects,Tubing ID in. 0.125,Material Acetal
EW-06363-57
In stock.
$88.00 USD/PACKAGE
Qty:
Representative photo only Hose barb
Hose barb
Product Type Miniature Quick-Disconnects,Tubing ID in. 0.0625,Material Acetal
EW-06363-54
In stock.
$20.50 USD/PACKAGE of 10
Qty:
EW-06363-56 Hose barb
Hose barb
Product Type Miniature Quick-Disconnects,Tubing ID in. 0.125,Material Acetal
EW-06363-56
In stock.
$18.00 USD/PACKAGE of 10
Qty:
Representative photo only Hose barb
Hose barb
Product Type Miniature Quick-Disconnects,Tubing ID in. 0.125,Material Polypropylene
EW-06363-64
In stock.
$22.00 USD/PACKAGE of 10
Qty:
Representative photo only Hose barb
Hose barb
Product Type Miniature Quick-Disconnects,Tubing ID in. 0.0625,Material Polypropylene
EW-06363-62
In stock.
$22.00 USD/PACKAGE of 10
Qty:
Representative photo only Miniature Quick Disconnect Hose Barb Female Straight through 1 8 ID
Miniature Quick-Disconnect Hose Barb (Female); Straight-through, 1/8" ID
Product Type Miniature Quick-Disconnects,Tubing ID in. 0.125,Material Polycarbonate
EW-06363-86
Usually ships in 13 days.
$36.00 USD/PACKAGE
Qty:
7 results (showing 1 - 7 )
Per Page 
Home  >  Fittings > Quick-Disconnect Fittings > Miniature Quick-Disconnect Fittings > Hose Barbs (female fitting halves)
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title