Home  >  Pressure & Vacuum Instruments > Presses > Hydraulic Press Test Systems, Manual
Hydraulic Press Test Systems, Manual
Hydraulic Press Test Systems, Manual
  • Test the physical limits of your sample—systems are ideal for a wide range of applications including material research and quality control
  • Compact design efficiently uses your laboratory bench space
  • Cast-iron base and bolsters with two threaded columns provide easy daylight adjustment
These heavy-duty systems handle a wide range of laboratory quality and performance applications such as compression, flexural, or shear strength; flow and color dispersion; and material research using ceramics, composites, construction materials, resins, and more. Safety features include a 10-gauge steel shield with see-through, shatterproof polycarbonate access door. The base has holes for permanent bench mounting and for optional permanent bench mounting.
Manual Hydraulic Press: Pressure gauge indicates both applied force and maximum pressure. Handle has cushioned comfort grip.
 Product Detail
Sort
 Availability
 Price
Representative photo only Hydraulic Press Test System Manual
Hydraulic Press Test System, Manual
Product Type Manual Hydraulic Press Test Systems
EW-59620-00
Usually ships in 9 days.
$2,750.00 USD/EACH
Qty:
1 results (showing 1 - 1 )
Per Page 
Home  >  Pressure & Vacuum Instruments > Presses > Hydraulic Press Test Systems, Manual
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title