United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Brand:
Cole-Parmer
(1)
Product Type:
Chemical Reagents
(14)
Chemical Bio-Reagents
(1)
Price Range:
$0 to $100
(8)
$100 to $299
(5)
$300 to $499
(1)
[+]
Show more
(1)
$500 to $799
(1)
[-]
Show less
Availability:
In Stock
(14)
Form:
Liquid
(12)
Quantity (mL):
500
(5)
4000
(3)
1000
(1)
[+]
Show more
(3)
100
(1)
25
(1)
473
(1)
[-]
Show less
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Chemicals, Reagents, & Lubricants
>
Chemicals
>
Chemicals Ha to Hy
>
Homatro to Hydroq
> Hydrogen Peroxide
Hydrogen Peroxide
Choose from our comprehensive line of Hydrogen Peroxide and other Homatro to Hydroq products.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Hydrogen Peroxide (3% w/v/USP) (4 liter) PO
Product Type Chemical Reagents,Purity grade USP,Form Liquid
EW-88012-24
In stock.
$289.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen Peroxide (50%, Stabilized/Certified) (500mL)
Product Type Chemical Reagents,Purity grade Certified,Form Liquid
EW-88012-08
In stock.
$187.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen Peroxide (30%/Certified ACS) (100mL)
Product Type Chemical Reagents,Purity grade Certified ACS,Form Liquid
EW-88012-10
In stock.
$73.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen Peroxide (30%/Certified ACS) (500mL)
Product Type Chemical Reagents,Purity grade Certified ACS,Form Liquid
EW-88012-12
In stock.
$157.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen peroxide, 35 wt.% solution in water, stabilized p.a. (1l)
Product Type Chemical Reagents,Form Liquid,Hazard symbol Corrosive
EW-88216-33
In stock.
$81.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen Peroxide (34-37%/Technical) (500mL)
Product Type Chemical Reagents,Purity grade Technical,Form Liquid
EW-88012-18
In stock.
$165.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen Peroxide (30% in Water) (500g)
Product Type Chemical Bio-Reagents,Hazard symbol Oxidizing, Corrosive
EW-88061-24
In stock.
$71.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen peroxide, 35 wt.% solution in water, stabilized p.a. (500ml)
Product Type Chemical Reagents,Form Liquid,Hazard symbol Corrosive
EW-88216-36
In stock.
$55.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen Perox 3% Reagent 473mL
Product Type Chemical Reagents,Purity grade Reagent,Form Liquid
EW-89000-86
Usually ships in 6 days.
$33.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen Peroxide (3% w/v/USP) (500mL)
Product Type Chemical Reagents,Purity grade USP,Form Liquid
EW-88012-22
In stock.
$88.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen Peroxide (30%/Certified ACS) (4 liter)
Product Type Chemical Reagents,Purity grade Certified ACS,Form Liquid
EW-88012-14
In stock.
$746.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen Peroxide (3% w/v/USP) (4 liter)
Product Type Chemical Reagents,Purity grade USP,Form Liquid
EW-88074-95
In stock.
$316.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen peroxide, reagent ACS (100g)
Product Type Chemical Reagents,Hazard symbol Corrosive
EW-88349-00
In stock.
$54.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen peroxide, reagent ACS (500g)
Product Type Chemical Reagents,Hazard symbol Corrosive
EW-88349-01
In stock.
$120.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
Hydrogen peroxide, 35 wt.% solution in water, stabilized p.a. (25ml)
Product Type Chemical Reagents,Form Liquid,Hazard symbol Corrosive
EW-88216-35
In stock.
$45.50 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hydrogen Peroxide
15
results (showing
1
-
15
)
Per Page
5
10
15
25
50
Home
>
Chemicals, Reagents, & Lubricants
>
Chemicals
>
Chemicals Ha to Hy
>
Homatro to Hydroq
> Hydrogen Peroxide
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title