United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Availability:
In Stock
(1)
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Mixers and Blenders
>
Mills
>
IKA Analytical Mills
> IKA Heavy-Duty Analytical Mill
IKA Heavy-Duty Analytical Mill
High-speed blade grinds samples in seconds
Grind dry and brittle sample volumes up to 250 mL
Cool samples with a recirculating bath or tap water
Mill is ideal for dried vegetation, soil samples, plastic granulates, frozen food samples, and more. Keep samples cool by circulating tap water or using a recirculating bath—the water-jacketed chamber accepts 3/8'' ID tubing via two barbed fittings.
Duty cycle ON/OFF (with cooling) is 7 minutes/10 minutes respectively. Automatic shut-off protection safeguards mill against overheating and overloading. Fan-cooled, universal-type 2/3-hp motor is completely isolated from the grinding chamber by a dust-tight seal. Mill is constructed of heavy-duty cast aluminum with enameled finish; cutting blade and internal chamber are 304 SS. Order an additional grinding chamber to run back-to-back tests—use one while cleaning and prepping the other. Note: Using liquids in the chamber may damage motor.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
IKA Heavy-Duty Analytical Mill, 115 VAC/60 Hz
Product Type Analytical Mills,Capacity 250 mL,Brand IKA
EW-04301-10
In stock.
$3,940.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
IKA
Heavy-Duty Analytical Mill
IKA Heavy-Duty Analytical Mill, 230 VAC/50 Hz
Product Type Analytical Mills,Capacity 250 mL,Brand IKA
EW-04301-15
Usually ships in 11 days.
$3,940.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
IKA Heavy-Duty Analytical Mill
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Mixers and Blenders
>
Mills
>
IKA Analytical Mills
> IKA Heavy-Duty Analytical Mill
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title