United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Availability:
In Stock
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Mixers and Blenders
>
Homogenizers
>
IKA Homogenizers
> IKA T-25 Digital High-Speed Homogenizer Systems
IKA T-25 Digital High-Speed Homogenizer Systems
Choose from seven generators with 0.3 to 2500 mL sample volumes
Systems can be used with tough solids or large volume samples
Generators are made of 316 stainless steel
Digital High-Speed Homogenizers
This system is ideal for batch homogenizing of cell tissues, emulsifying suspended solids,
and air pollution filter extraction. The key component is the rotor-stator generator. (Order rotorstator
generators separatelyfive volumes are available.) Shielded in a 316 stainless steel shaft,
the rotor acts as a centrifugal pump to recirculate the liquid and suspended solids through the
generator, where shear, impact, collision, and cavitation provide rapid homogenization.
Rotor-Stator Generators
Select a 316 SS shielded generator according to your sample volume, working solution, and
needed dispersion type. The standard-capacity generator is available in three volume ranges.
The large-capacity generator is available in both coarse (20 ìm) and fine (5 ìm) dispersion.
What''s included:
10-ft power cord (the 115 VAC model has a standard U.S. plug; the 230 VAC
model has a European plug).
,b>Kit includes: stand with an 111⁄2"L x 75⁄8"W base and 24"L stainless steel rod, beaker/test tube
holder, two movable clamps for positioning the shaft/generator and beaker/test tube holder,
and a tool kit for shaft assembly.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
IKA T 25 ULTRA-TURRAX High-Speed Homogenizer 115VAC-unit only
Product Type Homogenizer,Max sample size mL 2000,Speed rpm 500 to 25000
EW-04739-21
In stock.
$1,425.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
IKA T-25 Digital High-Speed Homogenizer Systems
IKA T 25 ULTRA-TURRAX High-Speed Homogenizer 220VAC-unit only
Product Type Homogenizer,Max sample size mL 2000,Speed rpm 500 to 25000
EW-04739-22
Call 1-888-358-4705 for availability.
$1,425.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
IKA T-25 Digital High-Speed Homogenizer Systems
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Mixers and Blenders
>
Homogenizers
>
IKA Homogenizers
> IKA T-25 Digital High-Speed Homogenizer Systems
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title