log in or register
Price Range:
Housing material:
PSF (2)
PPS (1)
Rotor material:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
I/P Easy-Load Pump Heads
I/P Easy-Load Pump Heads
  • Quick, simple tubing changes—tubing replacement and fluid changeover takes only seconds
  • Accept multiple tubing sizes for maximum flexibility
  • Automatic tubing retention eliminates user adjustments and makes setup easy—offers improved tubing life and improved performance
  • Fast tubing changes minimize downtime
  • Adjustable occlusion lets you optimize pressure and suction capabilities and tubing life by adjusting the “squeeze” on the tubing.
  • Five-screw mounting—ensures pump head is mounted securely to drive
The Easy-Load pump head combines an over-center cam with automatic tubing retention in a rugged, chemical-resistant housing. This patented design allows fast, easy tubing changes without removing the pump head from the drive. Built-in tubing guide ensures correct positioning of tubing to maximize performance and pump tubing life. Spring-loaded tubing retainers ensure tubing is secured automatically to promote long tubing life and optimize performance of each tubing size.
Select from polysulfone (PSF) housing with CRS or SS rotor, or polyphenylene sulfide (PPS) housing with SS rotor. PSF is economical, while PPS offers enhanced chemical resistance. SS rotors feature PTFE-sealed bearings for long life and reduced maintenance; CRS rotors have Buna N shielded bearings.
Dual-channel hardware allows two pump heads to operate on a single drive—order 77601-96 separately under "Accessories".
 Product Detail
Sort
 Availability
 Price
Representative photo only Masterflex I P Easy Load pump head PSF housing SS rotor
Masterflex I/P Easy-Load pump head, PSF housing/SS rotor.
Product Type Pump Head,Max flow rate LPM 13.0,Max flow rate GPM 3.5
EW-77601-10
In stock.
$660.00 USD/EACH
Qty:
EW-77601-60 Masterflex<small><sup>&reg;</sup></small> I/P<small><sup>&reg;</sup></small> Easy-Load<small><sup>&reg;</sup></small> pump head,  PPS housing/SS rotor.
Masterflex I/P Easy-Load pump head, PPS housing/SS rotor.
Product Type Pump Head,Max flow rate LPM 13.0,Max flow rate GPM 3.5
EW-77601-60
In stock.
$715.00 USD/EACH
Qty:
Representative photo only Masterflex I P Easy Load pump head PSF housing CRS rotor
Masterflex I/P Easy-Load pump head, PSF housing/CRS rotor.
Product Type Pump Head,Max flow rate LPM 13.0,Max flow rate GPM 3.5
EW-77601-00
In stock.
$510.00 USD/EACH
Qty:
3 results (showing 1 - 3 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title