United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Masterflex Peristaltic Pumps
>
Masterflex I/P Tubing Pumps
>
Masterflex I/P Drives
>
I/P Analog Variable-Speed Drives
>
I/P Precision Variable-Speed Modular Drives
> I/P Modular Drive
I/P Modular Drive
Flow rate: 0.12 to 13 LPM (0.03 to 3.5 GPM). Flow rate depends on drive rpm/tubing size.
6-ft (1.8-m) cable connects motor/controller. Place components where convenient.
Reversible 1/5-hp motor. Purge before pumping; pump fluid in either direction.
Maintain speed while turning drive on/off/reverse. Separate single-turn potentiometer, on/off/reverse inertia center switch, and power indicator.
6-ft (1.8-m) line cord. 115 VAC model: U.S. standard plug; 230 VAC model: IEC 320/CEE22 socket. (Specify country to receive the correct plug/cord set).
Accepts three different pump head types: Standard, EASY-LOAD
®
, or High-Performance pump heads
Combines all the features of the economy modular drive with a larger 1/5-hp motor. Add an extra pump head to drive when using silicone or C-FLEX
®
tubing.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Masterflex I/P variable speed modular drive, 20-650 rpm, 115V.
Product Type Masterflex I/P Drive,Max flow rate mL/min 19000,Min flow rate mL/min 120
EW-07591-00
In stock.
$1,675.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
I/P Modular Drive
Masterflex I/P Variable Speed Modular Drive, 20-650 RPM, 230V.
Product Type Masterflex I/P Drive,Max flow rate mL/min 17000,Min flow rate mL/min 120
EW-07591-07
In stock.
$1,675.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
I/P Modular Drive
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Masterflex Peristaltic Pumps
>
Masterflex I/P Tubing Pumps
>
Masterflex I/P Drives
>
I/P Analog Variable-Speed Drives
>
I/P Precision Variable-Speed Modular Drives
> I/P Modular Drive
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title