Home  >  Process Equipment & Instrumentation > Automation Control > Idec Operator Interfaces - Touch Screens
Idec Operator Interfaces - Touch Screens
Idec Operator Interfaces - Touch Screens
  • Operator interfaces are completely programmable and simple to use
  • Easily control PLCs
  • Touch screens offer easy control of automation and processes
  • Panel mount operator interfaces for convenience and ease of use
Operator interfaces are control units that enable you to control your PLC from the screen, either using a small control panel with character displays or the touch screen on the larger units. All the operator interfaces come with software that enable you to easily control you system.
Touch screens offer a perfect solution for your display need. Crisp images, combined with capable configuration tools, allow you to easily create a user-friendly and powerful system, enhancing your "Human Machine Interaction". All touch screens require 24 VDC and have communications ports. Touch screens are rated UL1604 for hazardous locations, NEMA 4X, and IP66.
All touch screens are programmable with software 65523-18. The symbols library of bitmap images has more than 4000 images in color and monochrome. WindO/I-NV2 also supports multiple screen handling using O/I link (up to 15 O/Is), and enables the user to uniquely tailor the program to suite the user’s needs. Order software and communication cables separately, at right.
Models 65523-00 and -02 have a bright LCD. Units can configure up to 100 pages of operational control and have 127 KB of memory. Display capabilities include bitmap and bar graph indicators, as well as a library of images and graphics. Models 65523-04 to -10 are full-function touch screens designed with a small profile. Models 65523-06 and 10 come with an exclusive technology called CC Click™- four large pushbuttons located below screen provide tactile and audio feedback and are easily programmed using software package. All four models have 2 MB of memory. Models 65523-12 and -14 support standard compact flash memory and have a built-in 10Base T Ethernet port and a parallel port. Both models have a field-replaceable backlight, and have 8 MB of memory.
 Product Detail
Sort
 Availability
 Price
Representative photo only Operator Interfaces Touch Screen Type HG2F SS22VF
Operator Interfaces - Touch Screen Type HG2F-SS22VF
Product Type Operator Interface,Display 256-color,Power VDC 24
EW-65523-08
Usually ships in 5 days.
$995.00 USD/EACH
Qty:
Representative photo only Operator Interfaces Touch Screen Type HG3F FT22TFW
Operator Interfaces - Touch Screen Type HG3F-FT22TFW
Product Type Operator Interface,Display 256-color backlit,Power VDC 24
EW-65523-12
Usually ships in 5 days.
$1,995.00 USD/EACH
Qty:
Representative photo only Operator Interfaces Touch Screens Type HG4F JT22TFW
Operator Interfaces - Touch Screens Type HG4F-JT22TFW
Product Type Operator Interface,Display 256-color TFT,Power VDC 24
EW-65523-14
Usually ships in 5 days.
$2,495.00 USD/EACH
Qty:
Representative photo only Operator Interfaces Touch Screen Type HG1B SB22WF
Operator Interfaces - Touch Screen Type HG1B-SB22WF
Product Type Operator Interface,Display 4" screen,Power VDC 24
EW-65523-02
Usually ships in 5 days.
$595.00 USD/EACH
Qty:
Representative photo only Operator Interfaces Touch Screen Type HG2F SS52VF
Operator Interfaces - Touch Screen Type HG2F-SS52VF
Product Type Operator Interface,Display 256-color with CC Click,Power VDC 24
EW-65523-10
Usually ships in 5 days.
$1,095.00 USD/EACH
Qty:
Representative photo only Operator Interfaces Touch Screen Type HG2F SB52VF
Operator Interfaces - Touch Screen Type HG2F-SB52VF
Product Type Operator Interface,Display monochrome with CC Click,Power VDC 24
EW-65523-06
Usually ships in 5 days.
$795.00 USD/EACH
Qty:
Representative photo only Operator Interfaces Touch Screen Type HG1B SB22JF
Operator Interfaces - Touch Screen Type HG1B-SB22JF
Product Type Operator Interface,Display 4" screen,Power VDC 24
EW-65523-00
Usually ships in 5 days.
$595.00 USD/EACH
Qty:
7 results (showing 1 - 7 )
Per Page 
Home  >  Process Equipment & Instrumentation > Automation Control > Idec Operator Interfaces - Touch Screens
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title