Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456

More Unique Lab & Industrial Supplies

 

Industrial Mixers

Industrial Mixers

Cole-Parmer offers an extensive line of overhead industrial mixers to suit mixing requirements from lab to scale-up. Our complete industrial mixer line includes heavy-duty mixers from hazardous duty to batch mixers. In addition to industrial mixers, we also offer a wide range of industrial mixer accessories including propellers, shafts, and stands.

Choosing a mixer for industrial use? Consider the following:

  • Container capacity
  • Liquid viscosity
  • Torque
  • Horsepower (hp)
  • Rotational speed (rpm) and diameter of mixing propeller
  • Duty cycle
  • Power supply

For more information on these variables, check out our Industrial Mixer Selection Guide for definitions and requirements. In addition to these requirements, the position of an overhead mixer in a vessel will create different mixing patterns and lead to different results. For more information on positioning an overhead industrial mixer in a vessel, see Industrial Mixer Positioning in a Cylindrical Vessel.

View our online catalog to see our full selection of products or contact our Application Specialists to help you find the right product for your needs.

Industrial Mixer details

Industrial mixers are used for mixing large quantities of material, or materials with higher viscosities and torque requirements. With the right mixer and propeller, these industrial units can mix up to 1500 gallons (5700 liters) of material with a viscosity of 1000 cps. Industrial mixers are available in electric or air-driven motors, with up to 1-hp motors. Mounting options include either a clamp (universal mounting) or 2" bung (for attaching directly to drums). Explosion proof and air drive mixers are available for locations where hazardous materials are used and sparks must be avoided. Our line of industrial mixers includes:

  • Bulk Container Mixers
  • Handle Bar Style Drum and Pail Mixers
  • Drum/Barrel Heavy-Duty Mixers
  • Drum/Barrel Top-to-Bottom Tube Mixers
  • Heavy-Duty Gear Drive Mixers
  • Lightnin Batch Industrial Mixers

Industrial Mixer Accessories to optimize performance

Cole-Parmer offers a number of accessories to optimize your mixer for industrial use. Our axial flow impellers feature a Superpitch design with a 1.5 pitch ratio for greater batch mixing capabilities. Marine impellers offer general purpose mixing applications. High-efficiency axial flow impellers are designed to provide 50% more mixing action and a true axial flow pattern. Folding impeller offer easy access through openings of 1-5/8" or more, providing full mixing capabilities once impeller opens. Folding impeller with integrated shaft provides full mixing with an unfolded diameter of 3 inches and a folded diameter of 1 inch, allowing you to fit the propeller and shaft through small bung holes. We also offer stainless steel shafts for industrial applications, ranging in length from 36 to 60 inches. For easier mounting and lowering of your industrial mixer with clamp, choose one of our steel industrial lift stands.

Need
Assistance?

Get help from
our Experts!
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price

Lightnin high-torque direct-drive mixer, variable-speed, offset clamp, 115 VAC

  • Product Type: Mixer
  • Impeller size: 3.8 in
  • Min speed: 90 rpm
EW-50323-20
Usually ships in 14 days or sooner.
$2,307.00 USD/EACH

Qty:

Drum/barrel heavy-duty mixer with C-clamp; 1/4 hp, 115/230 VAC

  • Product Type: Mixer
  • Impeller size: 4 in (fixed)
  • Motor: 0.25 hp
EW-04318-15
Usually ships in 12 days.
$908.00 USD/EACH

Qty:

Bung-Mount IBC Tote Mixer, 430 rpm, 1/2 HP; 115-230V

  • Product Type: Mixer
  • Drive type: Gear
  • Impeller size: 9 in
EW-50317-00
Usually ships in 10 days.
$1,889.00 USD/EACH

Qty:

Low-Profile Right-Angle IBC Tote Mixer, 350 rpm, 3/4 HP; 115-230V

  • Product Type: Mixer
  • Impeller size: 9 in diameter
  • Max speed: 350 rpm
EW-50316-08
Usually ships in 10 days.
$2,161.00 USD/EACH

Qty:

Drum/barrel tube mixer with air-driven motor

  • Product Type: Mixer
  • Max speed: 3000 rpm
  • Motor: 3/4 hp
EW-04316-22
In stock.
$1,461.00 USD/EACH

Qty:

Lightnin small batch fixed-speed mixer with 1/8-hp motor, 115 VAC

  • Product Type: Mixer
  • Impeller size: 3.4 in
  • Max speed: 1750 rpm
EW-04518-40
Available for order
or call 1-888-358-4705 to order
$1,893.00 USD/EACH

Qty:

Lightnin Heavy-Duty Gear Drive Mixer, TENV, 20-350rpm, 3/4" x 60", 10" impeller

  • Product Type: Mixer
  • Impeller size: 10 in
  • Min speed: 20 rpm
EW-50323-30
Usually ships in 14 days.
$3,019.00 USD/EACH

Qty:

Drum/barrel heavy-duty mixer with 2" NPT(M) mounting; 1/2 hp, 115/230 VAC

  • Product Type: Mixer
  • Impeller size: 3.75 in (folding)
  • Motor: 0.5 hp
EW-50500-10
Usually ships in 8 days.
$1,103.00 USD/EACH

Qty:

Lightnin heavy-duty, gear-drive mixer, XPRF

  • Product Type: Mixer
  • Impeller size: 10 in
  • Max speed: 350 rpm
EW-50310-46
Usually ships in 14 days.
$2,412.00 USD/EACH

Qty:

Bung-Mount IBC Tote Mixer, 430 rpm, 1 HP; 115-230V

  • Product Type: Mixer
  • Drive type: Gear
  • Impeller size: 9 in
EW-50317-10
Usually ships in 10 days.
$2,119.00 USD/EACH

Qty:

Lightnin high-torque direct-drive mixer, air-powered, offset clamp

  • Product Type: Mixer
  • Impeller size: 3.8 in
  • Min speed: 20 rpm
EW-50323-10
Usually ships in 10 days.
$1,511.00 USD/EACH

Qty:

Lightnin Heavy-Duty Gear Drive Mixer, TENV, 20-350rpm, 1440in-oz, 3/4" x 36" shaft

  • Product Type: Mixer
  • Impeller size: 8.8 in (dual)
  • Min speed: 20 rpm
EW-50310-04
Usually ships in 14 days.
$3,888.00 USD/EACH

Qty:

Lightnin Heavy-Duty Gear Drive Mixer, air, 20-350rpm, 1440in-oz, 3/4" x 60" shaft

  • Product Type: Mixer
  • Impeller size: 11.8 in
  • Min speed: 20 rpm
EW-50323-40
Usually ships in 12 days.
$2,654.00 USD/EACH

Qty:

Drum/barrel heavy-duty, air-drive mixer with 2" NPT(M) mounting

  • Product Type: Mixer
  • Impeller size: 3.75 in (folding)
  • Motor: 0.5 hp
EW-50500-20
Available for order
or call 1-888-358-4705 to order
$1,129.00 USD/EACH

Qty:

Heavy duty, gear-drive mixer; 1/2-hp, 36"L shaft, 8" impeller

  • Product Type: Mixer
  • Drive type: gear
  • Impeller size: 8 in
EW-50311-00
Available for order
or call 1-888-358-4705 to order
$1,889.00 USD/EACH

Qty:

94 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#