Home  >  Cooling Equipment > Ice Machines > Insulated Containers > Insulated Field Carrier
Insulated Field Carrier
Insulated Field Carrier
  • Handle for easy carrying - 1 cu ft capacity
Insulated field carrier has efficient, 2-1/4" thick polyurethane foam insulation core and rugged, double-wall polyethylene construction. Includes carrying handle, hinged lid, and adjustable latches. Inside dimensions are 16-5/8"L x 10-1/4"W x 10-3/4"H. NSF certified.
 Product Detail
Sort
 Availability
 Price
EW-03720-20 Insulated field carrier
Insulated field carrier
Product Type Ice Containers,Capacity cu ft 1
EW-03720-20
In stock.
$239.00 USD/EACH
Qty:
1 results (showing 1 - 1 )
Per Page 
Home  >  Cooling Equipment > Ice Machines > Insulated Containers > Insulated Field Carrier
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title