United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Industrial Equipment
>
Tools
>
Tool Kits
> Insulated Tool Kits
Insulated Tool Kits
Helps workers meet OSHA requirements
Rated for exposure up to 1000VAC and dielectrically tested at 10,000VAC
Meet or exceed ASTM 1505-07 and IEC 900 Standards for Insulated Hand Tools
Helps comply with OSHA 1910.333 (c)(2), and NFPA 70E 2004
These quality insulated tools are made with impact-resistant and flame-resistant material. Two-color insulation makes inspection easier which adds to the overall safety. If the yellow under layer is showing, the tool may no longer be insluated properly and should be considered for replacement and shall be removed from service.
Custom kits and individual tools available. Call our Application Specialists for more information.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
60 Piece Hot Box Insulated Tool Kit
Product Type Tool Sets,Qty/pk 60,Brand Salisbury
EW-91100-63
Usually ships in 4 days.
$3,075.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Insulated Tool Kits
30 Piece Deluxe Insulated Tool Kit
Product Type Tool Sets,Qty/pk 30,Brand Salisbury
EW-91100-53
Usually ships in 10 days.
$2,400.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Insulated Tool Kits
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Industrial Equipment
>
Tools
>
Tool Kits
> Insulated Tool Kits
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title