Irreversible High-Temperature 8- and 9-Point Vertical Labels / Indicators
Irreversible High-Temperature 8- and 9-Point Vertical Labels / Indicators
  • Reliable proof that the product was maintained at the correct temperature
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1.8°F below 212°F and ±1°F above 212°F. Shelf life: 1 year after date of purchase.
 Product Detail
Sort
 Availability
 Price
EW-08068-22 Eight-Point temperature indicating labels; temperature points 160-230°F (71-110°C), 25 pk
Eight-Point temperature indicating labels; temperature points 160-230F (71-110C), 25 pk
Product Type Temperature Labels,Dimensions " W 0.75,Dimensions " H 2
EW-08068-22
In stock.
$43.00 USD/PACKAGE of 25
Qty:
Representative photo only Eight Point temperature indicating labels temperature points 100 150F 37 65C 25 pk
Eight-Point temperature indicating labels; temperature points 100-150°F (37-65°C), 25 pk
Product Type Temperature Labels,Dimensions " W 0.75,Dimensions " H 2
EW-08068-20
In stock.
$43.00 USD/PACKAGE of 25
Qty:
Representative photo only Eight Point temperature indicating labels temperature points 240 310F 116 154C 25 pk
Eight-Point temperature indicating labels; temperature points 240-310°F (116-154°C), 25 pk
Product Type Temperature Labels,Dimensions " W 0.75,Dimensions " H 2
EW-08068-24
In stock.
$43.00 USD/PACKAGE of 25
Qty:
Representative photo only Eight Point temperature indicating labels temperature points 320 390F 160 199C 25 pk
Eight-Point temperature indicating labels; temperature points 320-390°F (160-199°C), 25 pk
Product Type Temperature Labels,Dimensions " W 0.75,Dimensions " H 2
EW-08068-26
In stock.
$43.00 USD/PACKAGE of 25
Qty:
Representative photo only Nine Point temperature indicating labels temperature points 400 500F 204 260C 25 pk
Nine-Point temperature indicating labels; temperature points 400-500°F (204-260°C), 25 pk
Product Type Temperature Labels,Dimensions " W 0.75,Dimensions " H 2
EW-08068-28
In stock.
$43.00 USD/PACKAGE of 25
Qty:
EW-08068-90 Eight- and nine-point indicator kit; contains five labels of each of the five types of indicators (Product #s 08068-20, 08068-22, 08068-24, 08068-26, 08068-28)
EW-08068-90
In stock.
$43.00 USD/PACKAGE of 25
Qty:
6 results (showing 1 - 6 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title