United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$100 to $299
(2)
$300 to $499
(4)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Furniture & Fume Hoods
>
Junction Boxes
> Junction Boxes -Type 316L Stainless Steel (Sizes ranging from: 6x6x4 to 16x14x6)
Junction Boxes -Type 316L Stainless Steel (Sizes ranging from: 6x6x4 to 16x14x6)
Provides protection for electrical components for indoor and outdoor environments
Foam-in-place gasket ensures perfect seal
Painted carbon steel RAL 7035 light gray, rated NEMA 12 (IP55), 3R, and 4 (IP66)
Stainless steel units available in 304SS or 316LSS, #4 brushed finish, rated NEMA 4X (IP66)
Galvanized panel included (WM wall-mount only)
Carbon steel enclosures have wall-mounting holes - SS enclosures have wall-mounting blind-nuts
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
16x14x6 Type 316L Junction Box, Stainless Steel
Product Type Junction Box Enclosures,Material Stainless Steel,Model JB161406H6
EW-30001-59
Usually ships in 7 days.
$395.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Junction Boxes -Type 316L Stainless Steel (Sizes ranging from: 6x6x4 to 16x14x6)
12x10x6 Type 316L Junction Box, Stainless Steel
Product Type Junction Box Enclosures,Material Stainless Steel,Model JB121006H6
EW-30001-56
Usually ships in 7 days.
$330.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Junction Boxes -Type 316L Stainless Steel (Sizes ranging from: 6x6x4 to 16x14x6)
14x12x6 Type 316L Junction Box, Stainless Steel
Product Type Junction Box Enclosures,Material Stainless Steel,Model JB141206H6
EW-30001-58
Usually ships in 6 days.
$381.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Junction Boxes -Type 316L Stainless Steel (Sizes ranging from: 6x6x4 to 16x14x6)
10x8x4 Type 316L Junction Box, Stainless Steel
Product Type Junction Box Enclosures,Material Stainless Steel,Model JB100804H6
EW-30001-55
Usually ships in 7 days.
$281.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Junction Boxes -Type 316L Stainless Steel (Sizes ranging from: 6x6x4 to 16x14x6)
12x12x6 Type 316L Junction Box, Stainless Steel
Product Type Junction Box Enclosures,Material Stainless Steel,Model JB121206H6
EW-30001-57
Usually ships in 7 days.
$353.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Junction Boxes -Type 316L Stainless Steel (Sizes ranging from: 6x6x4 to 16x14x6)
6x6x4 Type 316L Junction Box, Stainless Steel
Product Type Junction Box Enclosures,Material Stainless Steel,Model JB060604H6
EW-30001-54
Usually ships in 7 days.
$248.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Junction Boxes -Type 316L Stainless Steel (Sizes ranging from: 6x6x4 to 16x14x6)
6
results (showing
1
-
6
)
Per Page
5
10
15
25
50
Home
>
Furniture & Fume Hoods
>
Junction Boxes
> Junction Boxes -Type 316L Stainless Steel (Sizes ranging from: 6x6x4 to 16x14x6)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title