United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$100 to $299
(2)
$300 to $499
(1)
Rotor material:
Stainless steel
(2)
Cold-rolled steel
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Pump Heads
>
Masterflex L/S Pump Heads
>
L/S Easy-Load II Pump Heads
> L/S Easy-Load II Pump Heads for High-Performance Precision Tubing
L/S Easy-Load II Pump Heads for High-Performance Precision Tubing
Automatic retention holds tubing securely in place with no manual adjustmentjust load the tubing and start the pump
Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation.
Improved occlusion bed geometry lengthens tubing life.
Polyphenylene sulfide (PPS) housing with either stainless steel (SS) or cold-rolled steel (CRS) rotorchoose CRS for economy or SS for greater durability and chemical resistance.
Model 77201-62 with adjustable occlusion gives you the option of loosening occlusion for longer tubing life or increasing occlusion for higher pressure.
Each pump head for high-performance precision pump tubing accepts tubing sizes L/S
®
15, L/S
®
24, L/S
®
35, and L/S
®
36. Achieve flow rates of 1.7 to 2900 mL/min (1 to 600 rpm).
These pump heads accept the four high-performance precision tubing sizes. High-performance precision tubing has a thicker wall than precision tubing enabling better pressure generation, greater suction lift, and greater ability to pump viscous fluids.
What's included:
single-channel mounting hardware and a 15'' (38-cm) length of silicone pump tubing. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Masterflex L/S Easy-Load II pump head for high-performance precision tubing, fixed occlusion, SS rotor.
Product Type Pump Head,Rotor material Stainless steel,Housing material Polyphenylene sulfide
EW-77200-62
In stock.
$296.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Easy-Load II Pump Heads for High-Performance Precision Tubing
Masterflex L/S Easy-Load II pump head for high-performance precision tubing, adjustable occlusion, SS rotor.
Product Type Pump Head,Rotor material Stainless steel,Housing material Polyphenylene sulfide
EW-77201-62
In stock.
$338.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Easy-Load II Pump Heads for High-Performance Precision Tubing
Masterflex L/S Easy-Load II pump head for high-performance precision tubing, fixed occlusion, CRS rotor.
Product Type Pump Head,Rotor material Cold-rolled steel,Housing material Polyphenylene sulfide
EW-77200-52
In stock.
$228.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Easy-Load II Pump Heads for High-Performance Precision Tubing
3
results (showing
1
-
3
)
Per Page
5
10
15
25
50
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Pump Heads
>
Masterflex L/S Pump Heads
>
L/S Easy-Load II Pump Heads
> L/S Easy-Load II Pump Heads for High-Performance Precision Tubing
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title