United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$200 to $250
(1)
$250 to $299
(2)
Housing material:
Polysulfone
(2)
Polyphenylene sulfide
(1)
Rotor material:
Stainless steel
(2)
Cold-rolled steel
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Pump Heads
>
Masterflex L/S Pump Heads
>
L/S Easy-Load Pump Heads
> L/S Easy-Load Pump Heads for Precision Tubing
L/S Easy-Load Pump Heads for Precision Tubing
Ideal for fast tubing changes and reduced maintenance
Patented design and adjustable tubing retention allow for multiple tubing sizeschange tubing without removing head from drive.
Polysulfone (PSF) housing with either stainless steel (SS) or cold-rolled steel (CRS) rotorchoose CRS for economy or SS for greater durability and chemical resistance.
Model 07518-60 with polyphenylene sulfide (PPS) housing and SS rotor offers enhanced chemical resistance.
Each pump head for precision pump tubing accepts tubing sizes L/S
®
13, L/S
®
14, L/S
®
16, L/S
®
25, L/S
®
17, and L/S
®
18. Achieve flow rates of 0.06 to 2300 mL/min (1 to 600 rpm).
What''s included:
single-channel mounting hardware and a 15" (38-cm) length of silicone pump tubing.
Two-, three-, and four-channel mounting hardware is available separately. Stack pump heads up to the specified limits of your L/S
®
drive.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Masterflex L/S Easy-Load pump head for precision tubing; PSF housing, CRS rotor.
Product Type Pump Head,Rotor material Cold-rolled steel,Housing material Polysulfone
EW-07518-00
In stock.
$206.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Easy-Load Pump Heads for Precision Tubing
Masterflex L/S Easy-Load pump head for precision tubing; PSF housing, SS rotor.
Product Type Pump Head,Rotor material Stainless steel,Housing material Polysulfone
EW-07518-10
In stock.
$258.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S
Easy-Load Pump Heads for Precision Tubing
Masterflex L/S Easy-Load pump head for precision tubing; PPS housing, SS rotor.
Product Type Pump Head,Rotor material Stainless steel,Housing material Polyphenylene sulfide
EW-07518-60
In stock.
$285.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S
Easy-Load Pump Heads for Precision Tubing
3
results (showing
1
-
3
)
Per Page
5
10
15
25
50
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Pump Heads
>
Masterflex L/S Pump Heads
>
L/S Easy-Load Pump Heads
> L/S Easy-Load Pump Heads for Precision Tubing
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title