United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
No. of Pump heads accepted:
2
(2)
1
(2)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Masterflex Peristaltic Pumps
>
Masterflex L/S Tubing Pumps
>
Masterflex L/S Drives
>
L/S Analog Variable-Speed Drives
> L/S Economy Variable-Speed Console Drives
L/S Economy Variable-Speed Console Drives
An economical and versatile variable-speed drive with a wide flow range
Separate single-turn speed control and on/off switch with green power indicatormaintain speed setting while turning pump on/off.
The 1/20-hp unidirectional motor has soft start and back EMF for ±5% speed control.
Stackable painted steel housing is IP22 rated for protection from vertically falling water.
115 VAC models are UL and cUL listed; 230 VAC models are CE marked in compliance with European Community standards.
6-ft (1.8-m) line cord; 115 VAC: U.S. standard plug; 230 VAC: IEC 320/CEE22 socketpumps are shipped with cord/plug set specific to destination country.
Our lowest priced variable-speed drives are a perfect upgrade from a fixed-speed drive. These drives include a wider rpm range, ±5% speed regulation, and UL, cUL, and CE listings. For added versatility, these drives accept the L/S
®
pump head styles specified below.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Masterflex L/S economy variable-speed drive, 20 to 600 rpm, 115 VAC.
Product Type Pump Drive,Max flow rate mL/min 2900,Min flow rate mL/min 1.2
EW-07554-90
In stock.
$638.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Economy Variable-Speed Console Drives
Masterflex L/S economy variable-speed drive, 7 to 200 rpm, 115 VAC.
Product Type Pump Drive,Max flow rate mL/min 1130,Min flow rate mL/min 0.42
EW-07554-80
In stock.
$638.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Economy Variable-Speed Console Drives
Masterflex L/S economy variable-speed drive, 20 to 600 rpm, 230 VAC.
Product Type Pump Drive,Max flow rate mL/min 2900,Min flow rate mL/min 1.2
EW-07554-95
In stock.
$638.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Economy Variable-Speed Console Drives
Masterflex L/S economy variable-speed drive, 7 to 200 rpm, 230 VAC.
Product Type Pump Drive,Max flow rate mL/min 1130,Min flow rate mL/min 0.42
EW-07554-85
In stock.
$638.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Economy Variable-Speed Console Drives
4
results (showing
1
-
4
)
Per Page
5
10
15
25
50
Home
>
Masterflex Peristaltic Pumps
>
Masterflex L/S Tubing Pumps
>
Masterflex L/S Drives
>
L/S Analog Variable-Speed Drives
> L/S Economy Variable-Speed Console Drives
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title