United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
No. of Pump heads accepted:
2
(1)
4
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Masterflex Peristaltic Pumps
>
Masterflex L/S Tubing Pumps
>
Masterflex L/S Drives
>
L/S Analog Variable-Speed Drives
>
L/S Variable-Speed Modular Analog Drives
> L/S Precision Modular Drives with Bench Top Controller
L/S Precision Modular Drives with Bench Top Controller
Modular drives let you place motor and controller where convenient—ideal for limited spaces, hoods, or isolation chambers
Flow rate 0.06 to 3400 mL/min (depends on drive rpm and tubing size)
Three-digit LED display shows motor rpm—confirm speed setting at a glance
Membrane keypad for easy up-front control of speed, direction, and start/stop
Reversible motor; purge tubing before or after pumping; pump fluid in either direction
LED indicators confirm motor direction
Powerful motors let you stack 2 or 4 pump heads for increased flow rates and for multichannel applications
Separate speed and power (on/off) control; maintain speed setting while turning pump on/off
±0.25% speed control; meter and transfer fluids with accuracy and repeatability
Remote control capabilities via a DB9 female connector on back of drive: Speed control input 4 to 20 mA or 0 to 10 V; Pump direction (CW/CCW) via open collector or contact closure; Start/stop via open collector or contact closure
Splash-resistant ABS plastic controller housing is IP33 rated; drive housing is epoxy finished steel, IP33 rated
6-ft (1.8-m) cable connects drive and controller; use optional extension cable 07557-51 to extend to 15 ft (4.6 m)
L/S® modular drives feature reversible pumping and allow you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Masterflex L/S Precision Modular Drive with Bench Top Controller and Remote I/O; 6 to 600 rpm, 90 to 260 VAC
Product Type Pump Drive,Max flow rate mL/min 3400,Min flow rate mL/min 0.36
EW-07557-00
In stock.
$1,190.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Masterflex L/S Precision Modular Drive with Bench Top Controller and Remote I/O; 1 to 100 rpm, 90 to 260 VAC
Product Type Pump Drive,Max flow rate mL/min 580,Min flow rate mL/min 0.06
EW-07557-10
In stock.
$1,190.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Masterflex Peristaltic Pumps
>
Masterflex L/S Tubing Pumps
>
Masterflex L/S Drives
>
L/S Analog Variable-Speed Drives
>
L/S Variable-Speed Modular Analog Drives
> L/S Precision Modular Drives with Bench Top Controller
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title