United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
No. of Pump heads accepted:
2
(2)
4
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Masterflex Peristaltic Pumps
>
Masterflex L/S Tubing Pumps
>
Masterflex L/S Drives
>
L/S Analog Variable-Speed Drives
> L/S Precision Variable-Speed Console Drives
L/S Precision Variable-Speed Console Drives
Easy-to-use drives offer precise speed control and digital display of rpm—ideal for metering applications
Flow rate 0.06 to 3400 mL/min (depends on drive rpm and tubing size)
Three digit LED display shows motor rpmconfirm speed setting at a glance
Reversible motor—purge tubing before or after pumping; pump fluid in either direction
LED indicators confirm motor direction
Powerful motors let you stack 2 or 4 pump heads for increased flow rates and for multichannel applications
Separate speed and power (on/off) controlmaintain speed setting while turning pump on/off
Stackable, splash-resistant ABS plastic housing; IP33 rated
Remote control capabilities via a DB9 female connector on back of drive
Speed control input: 4 to 20 mA or 0 to 10 VDC
Pump direction (CW/CCW) via open collector or contact closure
Start/stop via open collector or contact closure
Membrane keypad for easy up-front control of speed, direction, and start/stop
These accurate analog console drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications. Remote speed control requires DB9 male connector 07595-45 or cable assembly 07595-47. Use remote connector for control of start/stop via contact closure or open collector (5 V TTL); or control start/stop via footswitch 77595-35. Order control accessories on facing page.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Masterflex L/S Precision Variable-Speed Drive with Remote Input; 6 to 600 rpm, 90 to 260 VAC.
Product Type Pump Drive,Max flow rate mL/min 3400,Min flow rate mL/min 0.36
EW-07528-10
In stock.
$1,120.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Precision Variable-Speed Console Drives
Masterflex L/S Precision Variable-Speed Drive with Remote Input; 1 to 100 rpm, 90 to 260 VAC.
Product Type Pump Drive,Max flow rate mL/min 580,Min flow rate mL/min 0.06
EW-07528-30
In stock.
$1,120.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Precision Variable-Speed Console Drives
Masterflex L/S Precision Variable-Speed Drive with Remote Input; 3 to 300 rpm, 90 to 260 VAC.
Product Type Pump Drive,Max flow rate mL/min 1700,Min flow rate mL/min 0.18
EW-07528-20
In stock.
$1,120.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Precision Variable-Speed Console Drives
3
results (showing
1
-
3
)
Per Page
5
10
15
25
50
Home
>
Masterflex Peristaltic Pumps
>
Masterflex L/S Tubing Pumps
>
Masterflex L/S Drives
>
L/S Analog Variable-Speed Drives
> L/S Precision Variable-Speed Console Drives
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title