United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Category:
Filter Papers
(330)
Membranes
(279)
Syringe Filters
(164)
Mesh Screen Discs, Sheets and Net Filters
(101)
Bottletop/Filter Units
(97)
Capsules and In-Line Filters
(93)
Vacuum Filtration Holders and Manifolds
(64)
Centrifugal Devices and Filters
(52)
In-Line and Syringe Filter Holders
(41)
Hollow Fiber Filters
(37)
Stirred Cells
(29)
Pressure Filtration
(24)
Brand:
Advantec
(545)
Whatman
(160)
Spectra/Mesh
(100)
[+]
Show more
(17)
Millipore
(98)
Cole-Parmer
(90)
Thermo Scientific Nalgene
(40)
Corning
(28)
MediaKap
(14)
Minntech
(14)
Autovial
(7)
Costar Spin-X
(6)
Labglass
(5)
MiniKap
(4)
Savillex
(3)
Millipak
(2)
Millipore Amicon
(2)
Masterflex
(1)
CultureGard
(1)
Vacu-Guard
(1)
Millipore Steripak
(1)
[-]
Show less
Product Type:
Filter Papers
(381)
Syringe Filters
(140)
Screen and Mesh Sheets/Discs
(100)
[+]
Show more
(17)
Mixed Cellulose Ester Membranes
(92)
Capsule Filters
(48)
PTFE Hydrophobic Membranes
(45)
Hollow-Fiber Filters
(33)
Vacuum Filtration Holders
(26)
Cellulose Acetate Membranes
(26)
Accessories for filter holders
(24)
Extraction Thimbles
(24)
PTFE Membranes
(22)
Nylon Membranes
(21)
Bottletop Filters
(21)
Gas/Liquid Filters, In-Line
(20)
Centrifugal Filters
(19)
Polyethersulfone Filter Units
(16)
Vacuum Filtration Systems
(14)
PTFE Hydrophilic Membranes
(12)
Cellulose Nitrate Filter Units
(11)
[-]
Show less
Price Range:
$0 to $100
(427)
$100 to $299
(660)
$300 to $499
(133)
[+]
Show more
(4)
$500 to $799
(51)
$800 to $1,099
(13)
$1,100 to $1,999
(18)
$2,000 and Over
(13)
[-]
Show less
Availability:
In Stock
(323)
Capacity (Liters):
0 to 1
(4)
1 to 5
(2)
10 to 50
(6)
Capacity (mL):
0 - 20
(2)
80 - 100
(20)
100 - 500
(38)
[+]
Show more
(4)
500 - 1000
(15)
2000
(1)
4000
(1)
5000
(2)
[-]
Show less
Diameter (mm):
50 and Above
(186)
Material:
Nylon
(73)
Mixed Cellulose Ester
(54)
PTFE
(50)
[+]
Show more
(17)
Hydrophobic PTFE
(36)
Polypropylene
(32)
Cellulose Acetate
(32)
Stainless Steel
(28)
Hydrophobic PTFE with PP support
(24)
Hydrophilic PTFE
(24)
Mixed Cellulose Ester, White Membrane, individually packaged
(18)
PEEK
(16)
Glass
(15)
Polysulfone
(15)
Silver
(15)
Cellulose Nitrate
(13)
304 Stainless Steel
(12)
316 Stainless Steel
(12)
Polycarbonate
(11)
Polyethersulfone
(10)
Sintered Glass
(8)
[-]
Show less
Pore size (micron):
0.45
(70)
0.20
(51)
0.2
(43)
[+]
Show more
(17)
20-25
(24)
2.5
(22)
5 to 10
(21)
8
(18)
< 5
(14)
1.2
(13)
2.7
(12)
0.22
(10)
1.6
(10)
1.5
(9)
11
(8)
0.6
(7)
> 10
(7)
2 to 10
(7)
0.4
(6)
0.7
(6)
5
(6)
[-]
Show less
Membrane material:
Polyethersulfone (PES)
(63)
Cellulose Acetate
(27)
Nylon
(24)
[+]
Show more
(17)
PTFE Hydrophobic
(14)
Polytetrafluoroethylene (PTFE)
(12)
Surfactant-Free Cellulose Acetate (SFCA)
(12)
PTFE
(12)
Polypropylene (PP)
(11)
Cellulose Nitrate
(10)
Mixed Cellulose Ester Hollow Fiber
(4)
Polyvinylidene fluoride (PVDF)
(4)
Quartz Microfiber
(4)
PTFE Hydrophilic
(4)
Mixed Cellulose Ester
(4)
PVDF
(4)
Regenerated Cellulose (RC)
(4)
Cellulose acetate (CA)
(4)
Regenerated Cellulose
(3)
Glass microfiber (GMF)
(3)
Borosilicate Glass
(2)
[-]
Show less
Housing material:
Polypropylene
(58)
Polystyrene
(38)
Acrylic
(29)
[+]
Show more
(1)
Polypropylene Upper; High-Impact Polystyrene Lower
(4)
[-]
Show less
Qty/bx:
50
(18)
100
(13)
20
(2)
[+]
Show more
(1)
25
(1)
[-]
Show less
Funnel size (mL):
300
(13)
500
(11)
1000
(10)
[+]
Show more
(5)
250
(5)
100
(4)
15
(4)
150
(4)
13/100
(1)
[-]
Show less
Diameter:
50 mm
(9)
70 mm
(8)
90 mm
(6)
[+]
Show more
(2)
25 mm
(3)
47 mm
(3)
[-]
Show less
Capacity (gal.):
1-5
(6)
Effective filtration area:
500 cm
2
(4)
550 cm
2
(4)
1150 cm
2
(4)
[+]
Show more
(17)
225 cm
2
(3)
1000 cm
2
(3)
2200 cm
2
(3)
900 cm
2
(3)
16 cm
2
(2)
1100 cm
2
(2)
450 cm
2
(2)
570 cm
2
(2)
4 cm
2
(1)
625 cm
2
(1)
1300 cm
2
(1)
380 cm
2
(1)
760 cm
2
(1)
1140 cm
2
(1)
600 cm
2
(1)
1200 cm
2
(1)
82000 m
2
(1)
[-]
Show less
Learn more:
Basics of Cartridge Filtration
Isolating Genomic DNA from Whole Blood
Bitter Brew
[-]
Show Less
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Laboratory Supplies & Consumables
>
Filtration
> Laboratory Filtration Products
Laboratory Filtration Products
Products for laboratory bench-scale batch applications
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Advantec Mixed Cellulose Ester Membranes; 47 mm dia, 0.45 µm; 100/pack
Product Type Mixed Cellulose Ester Membranes,Material Mixed Cellulose Ester,Flow rate water 0.045 L/min/cm
2
EW-06639-14
In stock.
$123.00 USD/PACKAGE of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Advantec Mixed Cellulose Ester Membranes
Advantec Polypropylene Filter Holder for 47-mm membranes
Product Type Polypropylene Filter Holders,Material Polypropylene,Filtration area 3.5 cm
2
EW-06623-22
In stock.
$58.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Advantec Polypropylene Holders
Cole-Parmer Nylon Nonsterile Syringe Filters; 0.45 µ pore size; 25 mm diameter
Product Type Syringe Filters,Housing material Acrylic,Color Natural
EW-02915-14
In stock.
$180.00 USD/BOX of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer Nylon Syringe Filters
Thermo Scientific Nalgene Disposable Sterile Filter Units; membrane, grided, CN; capacity, 150mL; pore size, 0.45
; 12/pack
Product Type Cellulose Nitrate Filter Units,Housing material Polypropylene Upper; High-Impact Polystyrene Lower,Color White
EW-06730-04
In stock.
$110.00 USD/DOZEN
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Thermo Scientific Nalgene
Disposable Analytical Filter Units and Filter Funnels
Cole-Parmer PTFE Nonsterile Syringe Filters; 0.2µ
Product Type Syringe Filters,Housing material Polypropylene,Color Natural
EW-02915-28
In stock.
$95.00 USD/BOX of 20
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer PTFE Syringe Filters
Cole-Parmer PTFE Nonsterile Syringe Filters; 0.45µ
Product Type Syringe Filters,Housing material Polypropylene,Color Natural
EW-02915-22
In stock.
$180.00 USD/BOX of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer PTFE Syringe Filters
Cole-Parmer PTFE Nonsterile Syringe Filters; 0.20
Product Type Syringe Filters,Housing material Polypropylene,Color Natural
EW-02915-20
In stock.
$180.00 USD/BOX of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
PTFE Filters
Thermo Scientific Polyethersulfone (PES) Filter Units; membrane; PES; capacity, 500mL; pore size, 0.20µ; 12/pack
Product Type Polyethersulfone Filter Units,Sterile Yes,Membrane material Polyethersulfone (PES)
EW-06730-43
In stock.
$210.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Thermo Scientific Polyethersulfone (PES) Filter Units and Bottletop Filters
Cole-Parmer PTFE Nonsterile Syringe Filters; pore size; 0.45µ
Product Type Syringe Filters,Housing material Polypropylene,Color Natural
EW-29550-10
In stock.
$135.00 USD/PACKAGE of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer PTFE Syringe Filters
Thermo Scientific Polyethersulfone (PES) Filter Units; 150 mL, 0.20 µm Pore Size
Product Type Polyethersulfone Filter Units,Sterile Yes,Membrane material Polyethersulfone (PES)
EW-06730-31
In stock.
$92.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Thermo Scientific Polyethersulfone (PES) Filter Units and Bottletop Filters
Labglass
Filtration Assembly, 47mm dia, 300 mL
Product Type Filtration Assemblies,Diameter mm 47,Qty/pk 1
EW-34509-00
In stock.
$185.00 USD/CASE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Labglass
Filtration Assemblies
Advantec Grade No. 5C Ashless Quantitative Filter Paper; 18.5 cm dia.; pack of 100
Product Type Filter Papers,Filter size 18.5 cm,Diameter 18.5 cm
EW-81051-54
In stock.
$114.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Advantec Grade No. 5C Ashless Filter Paper for Fine Retention
Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.45µm; absorbent pads not included; gridded surface; 1000/pack (packaged individually)
Product Type Mixed Cellulose Ester Membranes,Material Mixed Cellulose Ester, White Membrane, individually packaged,Diameter mm 47
EW-06644-31
In stock.
$355.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Advantec Sterile Mixed Cellulose Ester Filter Membranes
Disposable In-Line Gas/Liquid Filters; Nylon; gas efficiency, 93%; liquid efficiency, 25µ; 5/pack
Product Type Gas/Liquid Filters, In-Line,Housing material Nylon,Max pressure psi 125
EW-02909-10
In stock.
$145.00 USD/PACKAGE of 5
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Disposable In-Line Gas/Liquid Filters
Labglass
Filtration Assembly, 90mm dia, 1000 mL
Product Type Filtration Assemblies,Diameter mm 90,Qty/pk 1
EW-34509-10
In stock.
$590.00 USD/CASE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Labglass Filtration Assemblies
1311
results (showing
1
-
15
)
1
2
3
4
5
6
Next >
>>
Per Page
5
10
15
25
50
Home
>
Laboratory Supplies & Consumables
>
Filtration
> Laboratory Filtration Products
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title