log in or register
Price Range:
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Life Science > Chromatography Products > Gas Chromatography (GC) > Gas Generators > Laboratory Gas Generators for GC Applications
Laboratory Gas Generators for GC Applications
Laboratory Gas Generators for GC Applications
  • Modular and stackable units of hydrogen, nitrogen, and air gas generators—all controlled through one CPU—provide you the flexibility your lab needs!
  • Assemble in any combination—each delivers a different gas for your analytical application
  • CPU for control up to 32 units—can be remotely monitored and controlled via Ethernet/Internet
  • Integral operating log with maintenance tables, report alarms
  • Stack vertically to reduce footprint and save valuable lab space
Hydrogen Generators
  • Reliable, uninterrupted production, no downtime to change cylinders. Continuous output of >99.9995% pure hydrogen—can be used to supply both detectors and to supply carrier gas.
  • Integrated safety systems, which can be interfaced with the CPU or PCs for remote control
  • Removable water feed tank
Nitrogen Generators
  • capable of delivering 500 mL/min with purity > 99.9995%
  • available in a standard version and the HC version which ensures 100% removal of hydrocarbons (<0.1 ppm for applications with high-sensitivity detectors)
  • Ability to integrate with dedicated air compressor
Zero Air Generators
  • capable of supplying 3,000 m>/min, complete with dryer and catalytic reactor for hydrocarbon removal <0.1 ppm
  • high-efficiency Palladium-Platinum HC system for removal of HCTs < 0.1 ppm for an absolutely
    stable and disturbance-free baseline
  • Serves up to 8 FIDs/NPDs
 Product Detail
Sort
 Availability
 Price
Representative photo only Laboratory Hydrogen Generator 600mL min 115 230 VAC
EW-86440-03
Usually ships in 10 days.
$9,900.00 USD/EACH
Qty:
Representative photo only Nitrogen Gas Generator HC 500mL min 115VAC
EW-86440-13
Usually ships in 10 days.
$6,645.00 USD/EACH
Qty:
Representative photo only Laboratory Hydrogen Gas Generator 200 mL min 115 230 VAC
EW-86440-01
Call 1-888-358-4705 for availability.
$5,900.00 USD/EACH
Qty:
Representative photo only Nitrogen Gas Generator GC 500mL min 230VAC
EW-86440-12
Usually ships in 10 days.
$5,600.00 USD/EACH
Qty:
EW-86440-15 CPU for Lab Gas Generators
EW-86440-15
Usually ships in 10 days.
$750.00 USD/EACH
Qty:
Representative photo only Base for Lab Gas Generators
EW-86440-16
Usually ships in 10 days.
$40.50 USD/EACH
Qty:
Representative photo only Laboratory Air Generator GC 3 000 mL min 230 VAC
EW-86440-07
Usually ships in 10 days.
$4,500.00 USD/EACH
Qty:
Representative photo only Nitrogen Gas Generator GC 500mL min 115VAC
EW-86440-11
Usually ships in 10 days.
$5,600.00 USD/EACH
Qty:
Representative photo only Laboratory Hydrogen Gas Generator 400 mL min 115 230 VAC
EW-86440-02
Usually ships in 10 days.
$7,900.00 USD/EACH
Qty:
Representative photo only Laboratory Air Generator TOC 3 000 mL min 230 VAC
EW-86440-05
Usually ships in 10 days.
$5,650.00 USD/EACH
Qty:
Representative photo only Laboratory Air Generator GC 3 000 mL min 115 VAC
EW-86440-06
Usually ships in 10 days.
$4,500.00 USD/EACH
Qty:
Representative photo only Laboratory Air Generator TOC 3 000 mL min 115 VAC
EW-86440-04
Usually ships in 10 days.
$5,650.00 USD/EACH
Qty:
Representative photo only Nitrogen Gas Generator HC 500mL min 230VAC
EW-86440-14
Usually ships in 10 days.
$6,300.00 USD/EACH
Qty:
13 results (showing 1 - 13 )
Per Page 
Home  >  Life Science > Chromatography Products > Gas Chromatography (GC) > Gas Generators > Laboratory Gas Generators for GC Applications
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title