Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Water Purification
>
Water Distillation Systems
> Laboratory Water Still
Laboratory Water Still
Reliable, easy to use, and safe to clean
Features a sturdy metal stand for either wall or bench mounting
Operate directly from tap water
This versatile water still is perfect for general laboratory distillations. Features a waste water drain below the level of the still—connect your drain pipe from the boiler straight downward without troublesome bends that may slow the flow rate.
Boiler and high-efficiency condenser are both constructed of borosilicate glass to ensure the highest in distillate purity. Heater is chromium plated and provides protection from overheating. Condenser features screw thread connections and plastic 3⁄8" ID hose barb fittings—change tubing quickly and safely without worrying about using excessive force on the glass.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Laboratory Water Still
Product Type Water Purification,Capacity 4 liters/hr,ASTM Water Grade Type II
EW-99293-00
In stock.
$1,750.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Laboratory Water Still
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Water Purification
>
Water Distillation Systems
> Laboratory Water Still
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity