United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Category:
Laser Levels and Alignment Tools
(3)
Diode Laser Modules
(1)
Brand:
Cole-Parmer
(1)
Price Range:
$150 to $200
(1)
$200 to $250
(2)
$250 to $299
(1)
Availability:
In Stock
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
> Laser Products
Laser Products
Laser alignment equipment is vital to a highly functioning lab. Using laser alignment equipment ensures your laboratory tools are accurate and your results are useful. Though laboratories understand the importance of laser alignment, many can’t decide whether this is something that should be done in-house with available laser alignment equipment or should be contracted out. For most application in-house laser alignment equipment will be most useful. But for workplaces with much larger application a contracted service may be the way to go. It’s a delicate balance one constructive of cost, convenience and staff confidence levels. If you do decide to do laser readjustments in-house Cole-Parmer has an array of laser products that provide an easy and inexpensive method for leveling or locating position on your large or small construction projects. Our lasers operate at 650nm wavelength, provide between 1mW and 5mW of energy, and are classified as either class II or class III devices. Also included are instruments that have built in inclinometers for precisely measuring angles and those that have digital displays for easy to read output. If you have questions about laser alignment equipment contact one of our Technical Experts who will be glad to assist you.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Miniature Laser Alignment Kit with Crosshair Image; Includes Lens 41960-54, 50 to 125 ft Max Range
Class II
EW-41935-22
Usually ships in 11 days.
$289.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Miniature Laser Alignment Kits with Crosshair Image
Miniature Laser Alignment Kit with Dot Image; No Accessory Lens, 500 ft Max Range
Class IIIa,Model LTM2HKD
EW-41935-00
Usually ships in 6 days.
$219.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Miniature Laser Alignment Kits with Dot Image
Miniature Laser Alignment Kit with Dot Image; No Accessory Lens, 250 ft Max Range
Class II,Model LTM2HKD-CL2
EW-41935-02
Usually ships in 9 days.
$219.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Miniature Laser Alignment Kits with Dot Image
Cole-Parmer Industrial Laser Module, 250 ft Max Range
Class II,Brand Cole-Parmer
EW-41925-02
In stock.
$165.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer Industrial Laser Modules
4
results (showing
1
-
4
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
> Laser Products
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title