United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Flow, Level & Valves
>
Level Control
>
Flowline Echotouch Transmitters and Controllers
> Level Transmitter/Controller
Level Transmitter/Controller
Continuous noncontact level measurement—perfect for a wide range of applications
Intrinsically safe and sanitary versions available for the most demanding applications
Configure internal 10 amp relay as an alarm or an automatic fill/empty switch
Echotouch ultrasonic transmitters are unaffected by changing conductivity and dielectric values. Echo-Map™ filtration eliminates false echo signal returns. Ideal for dirty, coating, scaling, or corrosive liquids. Easily enter all process parameters and calibrations with the push-button, menu-driven display housed in each unit.
Ultrasonic transmitters emit a continuous series of sound waves that reflect from the targeted material. The round trip time of each pulse is directly proportional to the level. This value is then converted to a 4 to 20 mA signal for output to data acquisition devices, recorders, or remote displays. For remote display and control capabilities, purchase remote relay controller 43312-50 separately.
Transmitters/controllers feature rugged NEMA 4X enclosure with PVDF transducer. LED digital display indicates level in inches or meters.This noncontact transmitter/controller has the advantage of an extended measurement range of 0.5 to 24.5 ft. Use the built-in 10 amp relay to control a pump, valve, alarm, or other apparatus. LED display reads in either inches or meters.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Flowline Echotouch
Level Transmitter/Controller; 6" TO 24.5 FT, 10 AMP, SPDT, 250 VAC
Product Type Level Sensors,Power VAC 250,Power VDC 5
EW-43312-20
Usually ships in 7 days.
$950.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Level Transmitter/Controller
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Flow, Level & Valves
>
Level Control
>
Flowline Echotouch Transmitters and Controllers
> Level Transmitter/Controller
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title