log in or register
Availability:
Max psi:
150 (12)
2500 (4)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Liquid Level Sensors
Liquid Level Sensors
  • Detect liquids regardless of conductivity, color, or specific gravity
  • Economical and compact—built-in solid-state switching electronics
  • One-unit installation—simply screws into the top or side of your tank
These dependable, electro-optic level sensors contain no moving parts to clog or wear. Sensors offer ±1 mm repeatability and are available in a wide variety of materials to meet even the most demanding applications.
Installation is simple and quick through the tank top, bottom, or side. The switches can be used for low-, intermediate-, or high-liquid level detection. Solid state switching ensures dependability over a long service life. Order controller separately.
 Product Detail
Sort
 Availability
 Price
EW-32772-00 Electro-optic point-level sensor; PSF, wet sink, 1/4
Electro-optic point-level sensor; PSF, wet sink, 1/4" NPT(M)
Product Type Optic Point-Level Sensors,Max temperature 176F,Max psi 150
EW-32772-00
In stock.
$118.00 USD/EACH
Qty:
EW-32772-20 Electro-optic point-level sensor; PSF, wet sink, flanged
Electro-optic point-level sensor; PSF, wet sink, flanged
Product Type Optic Point-Level Sensors,Max temperature 176F,Max psi 150
EW-32772-20
Call 1-888-358-4705 for availability.
$123.00 USD/EACH
Qty:
EW-32772-08 Electro-optic point-level sensor; PSF, wet sink, 1/2
Electro-optic point-level sensor; PSF, wet sink, 1/2" straight thread
Product Type Optic Point-Level Sensors,Max temperature 176F,Max psi 150
EW-32772-08
Call 1-888-358-4705 for availability.
$118.00 USD/EACH
Qty:
Representative photo only Electro optic point level sensor PTFE dry sink 1 4 NPT M
Electro-optic point-level sensor; PTFE, dry sink, 1/4" NPT(M)
Product Type Optic Point-Level Sensors,Max temperature 176F,Max psi 150
EW-32772-18
Call 1-888-358-4705 for availability.
$272.00 USD/EACH
Qty:
EW-32772-16 Electro-optic point-level sensor; PTFE, wet sink, 1/4
Electro-optic point-level sensor; PTFE, wet sink, 1/4" NPT(M)
Product Type Optic Point-Level Sensors,Max temperature 176F,Max psi 150
EW-32772-16
Call 1-888-358-4705 for availability.
$272.00 USD/EACH
Qty:
Representative photo only Electro optic point level sensor PSF dry sink 1 2 straight thread
Electro-optic point-level sensor; PSF, dry sink, 1/2" straight thread
Product Type Optic Point-Level Sensors,Max temperature 176F,Max psi 150
EW-32772-10
In stock.
$118.00 USD/EACH
Qty:
Representative photo only Electro optic point level sensor PSF dry sink 1 4 NPT M
Electro-optic point-level sensor; PSF, dry sink, 1/4" NPT(M)
Product Type Optic Point-Level Sensors,Max temperature 176F,Max psi 150
EW-32772-02
Call 1-888-358-4705 for availability.
$118.00 USD/EACH
Qty:
Representative photo only Electro optic point level sensor nylon dry sink 1 4 NPT M
Electro-optic point-level sensor; nylon, dry sink, 1/4" NPT(M)
Product Type Optic Point-Level Sensors,Max temperature 176F,Max psi 150
EW-32772-06
Call 1-888-358-4705 for availability.
$118.00 USD/EACH
Qty:
Representative photo only Electro optic point level sensor nylon wet sink 1 4 NPT M
Electro-optic point-level sensor; nylon, wet sink, 1/4" NPT(M)
Product Type Optic Point-Level Sensors,Max temperature 176F,Max psi 150
EW-32772-04
Call 1-888-358-4705 for availability.
$118.00 USD/EACH
Qty:
Representative photo only Electro optic point level sensor zinc nickel plated carbon steel dry sink 1 2 NPT M
Electro-optic point-level sensor; zinc/nickel-plated carbon steel, dry sink, 1/2" NPT(M)
Product Type Optic Point-Level Sensors,Max temperature 212F,Max psi 2500
EW-32772-26
Call 1-888-358-4705 for availability.
$191.00 USD/EACH
Qty:
Representative photo only Electro optic point level sensor PSF dry sink flanged
Electro-optic point-level sensor; PSF, dry sink, flanged
Product Type Optic Point-Level Sensors,Max temperature 176F,Max psi 150
EW-32772-22
Call 1-888-358-4705 for availability.
$123.00 USD/EACH
Qty:
EW-32772-30 Electro-optic point-level sensor; nickel-plated carbon steel/fused glass, wet sink, 1/2
Electro-optic point-level sensor; nickel-plated carbon steel/fused glass, wet sink, 1/2" NPT(M)
Product Type Optic Point-Level Sensors,Max temperature 212F,Max psi 2500
EW-32772-30
Usually ships in 7 days.
$145.00 USD/EACH
Qty:
Representative photo only Electro optic point level sensor Isoplast dry sink 3 8 NPT M
Electro-optic point-level sensor; Isoplast, dry sink, 3/8" NPT(M)
Product Type Optic Point-Level Sensors,Max temperature 212F,Max psi 150
EW-32772-14
Usually ships in 13 days.
$105.00 USD/EACH
Qty:
Representative photo only Electro optic point level sensor nickel plated carbon steel fused glass dry sink 1 2 NPT M
Electro-optic point-level sensor; nickel-plated carbon steel/fused glass, dry sink, 1/2" NPT(M)
Product Type Optic Point-Level Sensors,Max temperature 212F,Max psi 2500
EW-32772-32
Usually ships in 12 days.
$145.00 USD/EACH
Qty:
EW-32772-24 Electro-optic point-level sensor; zinc/nickel-plated carbon steel, wet sink, 1/2
Electro-optic point-level sensor; zinc/nickel-plated carbon steel, wet sink, 1/2" NPT(M)
Product Type Optic Point-Level Sensors,Max temperature 212F,Max psi 2500
EW-32772-24
Usually ships in 14 days.
$209.00 USD/EACH
Qty:
16 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title