United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Material Testing & Physical Testing
>
Melting Point
> METTLER TOLEDO Digital Melting Point Apparatus
METTLER TOLEDO Digital Melting Point Apparatus
Precision accuracy of ±0.5°C provides high reproducibility of results
Meet GLP requirements with optional printer
Digital display eliminates interpretation errors
This automatic melting point apparatus is ideal for determining sample purities for quality control, educational, and research and development applications. Unit measures the reflection of incident light in the crystalline state and the increase in light transmittance during the melting process. Operation is simpleinsert the test tube containing one to three mg of sample into the instrument, select the starting temperature and heating rate (user selectable between 0.1°C and 10°C per minute in steps of 0.1°C), and the visual and audible signals will indicate when the melting point is acheived.
The operator interface provides graphic display aids making set up and programming simple. The large LCD displays temperature readings to 0.1°C.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Capillary tubes, 1.3 to 1.5-mm dia., 150/pk.
Model 18552
EW-03014-62
In stock.
$103.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
METTLER TOLEDO Digital Melting Point Apparatus
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Material Testing & Physical Testing
>
Melting Point
> METTLER TOLEDO Digital Melting Point Apparatus
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title