Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$100 to $299
(1)
$300 to $499
(1)
$500 to $799
(1)
[+]
Show more
(1)
$1,100 to $1,999
(1)
[-]
Show less
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Nuclear Magnetic Resonance (NMR) Spectroscopy
> MNOVA NMR Predict Desktop Plug-In for Spectrometers
MNOVA NMR Predict Desktop Plug-In for Spectrometers
Easily predict 1-D and HSQC NMR spectra from a molecular structure
Verify your experimental spectra with prediction data
Keep your Mnova software up-to-date with one year of support and updates for free
Automatic solvent recognition and intelligent peak picking ensures optimal results
MNOVA NMRPredict Desktop Plug-in
generates of 1-D and Heteronuclear Single Quantum Coherence (HSQC) spectra from molecular structures and is ideal for the expert user looking for extensive or advanced functionality. Combined with the MNOVA NMR software, it provides a very simple interface to automatically analyze spectra. Features such as peak picking, integration, and multiplet analysis allow your experiments to run smoothly. The plug-in allows you to assign 1-D and 2-D spectra, make notes and reports in various journal formats, and paste molecular structures into your document. Further automation is allowed and access to pre-made scripts and processing templates is available; contact our Application Specialists for more details. The plug-in will operate in Windows®, Mac®, and various Linus environments.
The plug-in is available with either a perpetual or annual license. The perpetual license allows you to use the licensed software indefinitely. For the first year after purchase, the user is entitled to download any updates to the software and receive a full free year of technical support. Once the initial year ends, you can continue to purchase updates and support packages. The annual license will allow one year use of the licensed software, including updates and free technical support. Once the initial twelve months ends, the software will no longer function unless a new license is purchased.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
MNOVA NMR Predict Plug-In Annual License, for Academia and Government- NMR Software for processing, analysis, simulation and reporting
EW-83090-72
Usually ships in 4 days.
$140.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
MNOVA NMR Predict Desktop Plug-In for Spectrometers
MNOVA NMR Predict Plug-In Perpetual License, for Academia and Government- NMR Software for processing, analysis, simulation and reporting
EW-83090-73
Usually ships in 4 days.
$350.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
MNOVA NMR Predict Desktop Plug-In for Spectrometers
MNOVA NMR Predict Plug-In Annual License, for Industry- NMR Software for processing, analysis, simulation and reporting
EW-83090-76
Usually ships in 4 days.
$550.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
MNOVA NMR Predict Desktop Plug-In for Spectrometers
MNOVA NMR Predict Plug-In Perpetual License, for Industry- NMR Software for processing, analysis, simulation and reporting
EW-83090-77
Usually ships in 4 days.
$1,350.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
MNOVA NMR Predict Desktop Plug-In for Spectrometers
4
results (showing
1
-
4
)
Per Page
5
10
15
25
50
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Nuclear Magnetic Resonance (NMR) Spectroscopy
> MNOVA NMR Predict Desktop Plug-In for Spectrometers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity