United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$100 to $299
(1)
$300 to $499
(1)
$500 to $799
(1)
[+]
Show more
(1)
$1,100 to $1,999
(1)
[-]
Show less
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Nuclear Magnetic Resonance (NMR) Spectroscopy
> MNOVA Software packages for NMR Spectrometers
MNOVA Software packages for NMR Spectrometers
Easily predict 1-D and HSQC NMR spectra from a molecular structure
Verify your experimental spectra with prediction data
Keep your Mnova software up-to-date with one year of support and updates for free
Automatic solvent recognition and intelligent peak picking ensures optimal results
A traditional NMR spectrometer (nuclear magnetic resonance) can cost anywhere from hundreds of thousands of dollars, to millions of dollars. By contrast the NMR spectrometer offered through Cole-Parmer consists of one instrument: The world’s first affordable, miniature, easy-to-install and use, yet fully functional FT-NMR spectrometer—the picoSpin 45L. Cole-Parmer is proud to introduce the picoSpin 45L as a revolutionary instrument that greatly expands the availability of a NMR spectrometer. Unlike traditional NMR spectrometers, the picoSpin 45L’s compact size makes it the smallest true FT-NMR spectrometer available. Weighing in at only 10.5 lbs, this solid-state piece of electronics has no moving parts and fits easy into most laboratory settings. However, don’t let its small size fool you. The picoSpin-45L is a complete 45 MHz pulse NMR spectrometer, making it fully capable of monitoring reactions in your lab, pre-screening your samples, monitoring your reaction progress, or supporting many of the other applications for which NMR is known. As a fully functional FT-NMR instrument, it can not only help you identify detailed structural information surrounding chemical compounds, it can also be used for far-ranging applications. If you have a question about the NMR spectrometer offered by Cole-Parmer contact a Technical Expert today.
MNOVA NMR Software License
provides NMR processing, analysis and reporting for 1-D and 2-D NMR spectra right at your fingertips. It is ideal for both the expert user requiring advanced functionality, and for organic or synthetic chemists focused on routine processing with maximum productivity. Using the algorithms built into the software, you will be able to create synthesized spectra using only the peaks of your interest.
Easily transfer your data into report or patent formats. The MNOVA NMR software is formatted much like Powerpoint®, allowing a multipage interface and annotations with both text and graphics. MNOVA NMR software will operate in Windows®, Mac®, and various Linus environments.
The software package is available with either a perpetual or annual license. The perpetual license allows you to use the licensed software indefinitely. For the first year after purchase, the user is entitled to download any updates to the software and receive a full free year of technical support. Once the initial year ends, you can continue to purchase updates and support packages. The annual license will allow one year use of the licensed software, including updates and free technical support. Once the initial twelve months ends, the software will no longer function unless a new license is purchased.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
MNOVA Annual License for Industry, NMR Software for processing, analysis, simulatino and reporting
EW-83090-74
Usually ships in 4 days.
$650.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
MNOVA Software packages for NMR Spectrometers
MNOVA Perpetual License, for Industry- NMR Software for processing, analysis, simulation and reporting
EW-83090-75
Usually ships in 4 days.
$1,600.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
MNOVA Software packages for NMR Spectrometers
MNOVA Perpetual License, for Academia and Government- NMR Software for processing, analysis, simulation and reporting
EW-83090-71
Usually ships in 4 days.
$400.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
MNOVA Software packages for NMR Spectrometers
MNOVA Annual License, for Academia and Government- NMR Software for processing, analysis, simulation and reporting
EW-83090-70
Usually ships in 4 days.
$150.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
MNOVA Software packages for NMR Spectrometers
4
results (showing
1
-
4
)
Per Page
5
10
15
25
50
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Nuclear Magnetic Resonance (NMR) Spectroscopy
> MNOVA Software packages for NMR Spectrometers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title