United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
>
Multiparameter Meters
>
Handheld Multiparameter Meters
> MYRON L Waterproof 6Psi Multiparameter Meter
MYRON L Waterproof 6Psi Multiparameter Meter
IP67-rated Valox
®
housingsubmersible down to 3 ft (0.9 m)
Four-digit LCD reads to 9999 µS/ppm
6Psi reads pH, ORP, conductivity, TDS, resistivity, free chlorine, LSI and water hardness
These meters are simple and easy to usejust add your sample to the built-in cell cup, push the parameter key, and take a reading. Autoranging function automatically selects from five conductivity or TDS ranges.
Factory-calibrated meters have conductivity-to-TDS conversion curves for NaCl, 442, and KCl standards stored in memory. You can also calibrate using your own calibration values. Automatic temperature compensation with adjustable temperature coefficients up to 9.99%/°C provides accurate and precise measurements. Easily store and recall up to 100 measurements with time-and-date stamp. Nonvolatile memory retains all data and can be downloaded via bluDock
accessory ordered separately.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
MYRON L Waterproof 6Psi Multiparameter Meter
Product Type Multiparameter Meter,Meter style Portable,Dimensions " W 2-3/4
EW-19401-22
In stock.
$1,053.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
MYRON L Waterproof 6Psi Multiparameter Meter
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
>
Multiparameter Meters
>
Handheld Multiparameter Meters
> MYRON L Waterproof 6Psi Multiparameter Meter
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title