Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Masterflex Diss-Washer Mobile Wash Station
Masterflex Diss-Washer Mobile Wash Station
Masterflex Diss-Washer Mobile Wash Station
  • DISS-WASHER offers 300° spray angle at 20 psi. Clean your dissolution tester in minutes using less DI water.
  • Compact DISS-WASHER system rests on a 39-1/4"L x 22-1/2"W x 47-1/4"H stainless steel cart. DISS-WASHER moves easily throughout your lab.
  • System meets TOC sanitary standards for dissolution testing. Eliminates cross-contamination with organic carbons.
  • Eliminates vessel breakage or excessive water on your laboratory floor. Increases safety in your lab; never move your dissolution vessels for cleaning!
  • Includes in-house Installation Qualification (IQ) and Operational Qualification (OQ) documentation package. Save money by personally documenting your application procedure.
Add a MASTERFLEX DISS-WASHER to your dissolution tester cleaning process and eliminate the need to actually move the glass vessels to your sink for washing. Simply roll the DISS-WASHER up to the dissolution station, place the washhead over the vessels, and apply the foot pedal. The DISS-WASHER will drain away used media and wash the vessel with a 20 psi, 300° spray angle. Within minutes you are ready for your next dissolution protocol!
 Product Detail
Sort
 Availability
 Price
Thallium Standard 1 mL 1 mg Tl 1000 ppm Tl 120 mL  (Representative photo only)
EW-86992-46
Usually ships in 10 days.
$30.00 USD/EACH
Qty:
See all Thallium
Thallium AA Std 1000 ppm Tl Metal in Nitric 500mL (Representative photo only)
EW-86997-64
Usually ships in 12 days.
$71.00 USD/EACH
Qty:
See all Thallium
Thallium ICP Standard 1000 ppm 500ML (Representative photo only)
Thallium ICP Standard 1000 ppm 500ML
Product Type Chemical ICP Standards
EW-86997-66
Usually ships in 12 days.
$127.00 USD/EACH
Qty:
See all Thallium
Thallium Plasma Grade Reference Standard 1 mL 1 mg Tl 1000 ppm Tl 120 mL  (Representative photo only)
Thallium Plasma Grade Reference Standard, 1 mL = 1 mg Tl, 1000 ppm Tl (120 mL)
Purity grade Plasma Grade,Product Type Chemical ICP Standards
EW-86992-47
Usually ships in 12 days.
$66.00 USD/EACH
Qty:
See all Thallium
Thallium Standard 1 mL 1 mg Tl 1000 ppm Tl 500 mL  (Representative photo only)
EW-86976-80
Available for order
or call 1-888-358-4705 to order
$67.00 USD/EACH
Qty:
See all Thallium
Thallium i fluoride 99 5g (Representative photo only)
Thallium(i)fluoride 99% 5g
Product Type Chemical Reagents
EW-88286-23
In stock.
$41.00 USD/EACH
Qty:
See all Thallium
Thallium AA Std 1000 ppm Tl Metal in Nitric 100mL (Representative photo only)
EW-86997-63
Usually ships in 12 days.
$30.00 USD/EACH
Qty:
See all Thallium
Thallium ICP Standard 1000 ppm 100ML (Representative photo only)
Thallium ICP Standard 1000 ppm 100ML
Product Type Chemical ICP Standards
EW-86997-65
Usually ships in 12 days.
$104.00 USD/EACH
Qty:
See all Thallium
Thallium Plasma Grade Reference Standard 1 mL 10 mg Tl 10 000 ppm Tl 120 mL  (Representative photo only)
Thallium Plasma Grade Reference Standard, 1 mL = 10 mg Tl, 10,000 ppm Tl (120 mL)
Purity grade Plasma Grade,Product Type Chemical ICP Standards
EW-86992-48
Usually ships in 10 days.
$104.00 USD/EACH
Qty:
See all Thallium
9 results (showing 1 - 9 )
Per Page 
Home  >  Masterflex Diss-Washer Mobile Wash Station