United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Samplers
>
Water and Industrial Liquid Sampling
>
Peristaltic and Vacuum Fluid Sampling Pumps
>
Masterflex Fluid Sampling Pumps
> Masterflex E/S Composite Sampler
Masterflex E/S Composite Sampler
Program operation to meet EPA sampling requirements under EPA 833-B92.001 CFR40 Part 122.21 and 122.26
Controls can wait up to 90 days for a remote signal
Use one of the six preset programs or customize up to five for your specific needs
Adjustable back-lit LCD and glow-in-the-dark keypad help you to read the control panel in low-light environments
Slim design makes it easy to pass through standard 18-inch diameter manhole
When case is closed and latched, sampler floats in water for up to 30 minutes!
IP56-rated control panel protects against inclement weather and splashing water
Rechargeable battery makes it ideal for field applications
The versatile Masterflex
®
E/S
TM
composite sampler can collect a 9.0-liter sample while running unattendedideal for a variety of environmental field applications. The highly visible yellow housing makes it easy to spot and recover in the field.
The programmable control panel lets you set the sample size, sampling time, and delayed start. Start/stop and standby functions can be programmed for convenient remote control operation. The standby function delays the sampling up to 167 hours, 59 minutesyou don’t have to be there when the sampling begins.
The sampler lifts water, slurries, and liquid waste up to 25 feet, and logs 300 sampling events. The reversible flow function purges the tubing and removes material from the inlet. In-line sensors ensure volume accuracy and prevent the carboy from overfilling. The flow rate depends on the tubing you use.
The sampler can be powered by the built-in 12 VDC battery, or by the universal converter for 115 or 230 VAC operation. The converter can also recharge the 12 VDC battery. The battery will run the motor for three hours before it needs recharging. The LCD alerts you at startup when the internal battery needs recharging. Order the automotive power adapter separately below to run off of a DC power supply. Unit measures 13-1/2"W x 17-1/2"H x 11"D.
What's included:
a Masterflex
®
L/S
®
Easy-Load
®
pump head, a 9-liter HDPE carboy, 12 VDC rechargeable battery, 115/230 VAC power converter/battery recharger, quick-disconnect fittings for inlet and outlet connections, and 25-ft of L/S
®
peroxide-cured silicone tubing.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Masterflex E/S
Composite Sampler
Brand Masterflex
EW-07580-00
Call 1-888-358-4705 for availability.
$3,055.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Masterflex E/S Composite Sampler
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Samplers
>
Water and Industrial Liquid Sampling
>
Peristaltic and Vacuum Fluid Sampling Pumps
>
Masterflex Fluid Sampling Pumps
> Masterflex E/S Composite Sampler
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title