United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Masterflex Peristaltic Pumps
>
Masterflex I/P Tubing Pumps
>
Masterflex I/P Drives
>
I/P Air-Powered and Hazardous-Duty Drives
> Masterflex I/P Variable-Speed Hazardous-Duty Drives
Masterflex I/P Variable-Speed Hazardous-Duty Drives
Use either L/S
®
or I/P
®
pump head on drive. L/S flow rate: 0.6 to 2500 mL/min; I/P flow rate: 60 to 8800 mL/min. Flow rate depends on drive rpm and tubing size.
115 VAC motor: UL-listed for Class 1, Group D, Division 1 hazardous locations; 230/400 VAC motor: EEx-d IIBT4 rating. Use in areas where flammable gases/vapors are present.
Fully mechanical speed control—adjust with lever then lock setting. Mechanical control is inherently safer from sparking than electrical control.
230/400 VAC model complies with CE regulations. Conforms to design/manufacturing standards of European community.
Power switch and line cord are not included. Safely connect power using your own protected connections.
Drives accept ten different pump head types. L/S Pump Heads: Standard, EASY-LOAD
®
, EASY-LOAD
®
II, QUICK LOAD
®
, High-Performance, Cartridge, PTFE-Tubing, PTFE-Diaphragm; I/P Pump Heads: Standard, and EASY-LOAD.
Choose one of these variable-speed hazardous-duty drives for hazardous locations where special electrical precautions are necessary.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Masterflex I/P variable-speed hazardous-duty drive, 10-430 rpm, 115V.
Product Type Masterflex I/P Drive,Max flow rate mL/min 11500,Min flow rate mL/min 60
EW-07583-50
Usually ships in 14 days or sooner.
$2,570.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Masterflex I/P Variable-Speed Hazardous-Duty Drives
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Masterflex Peristaltic Pumps
>
Masterflex I/P Tubing Pumps
>
Masterflex I/P Drives
>
I/P Air-Powered and Hazardous-Duty Drives
> Masterflex I/P Variable-Speed Hazardous-Duty Drives
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title