Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Tubing and Pump Accessories
> Masterflex Linkable Instrument Control Software (WinLIN)
Masterflex Linkable Instrument Control Software (WinLIN)
Control up to 25 Masterflex
®
L/S
®
07551-series drives or Cole-Parmer Servodyne mixers directly from your PC
Interface the 07551-series computer-compatible drives via either RS-232 or USB connection
Customize the software to meet your application needs: dispensing, mixing/diluting, flow proportioning, and single/multiple slope gradients; program each pump to automate up to 50 steps.
Balance interface allows you to dispense off of a balance and also features weight-ratio dispensing; use of multiple balances is enabled with multiple COM ports on your PC
Logging options let you log at specific intervals, at the end of each block, or at the end of each run
Complies with 21 CFR Part 11: password protection and authorized user list, file encryption permits authorized user access only.
Windows
®
-based software has familiar pull down menus and mouse-selectable icons.
For PC interface via RS-232 port and for daisy-chaining drive to drive use DB9 M/F cable 22050-54; for PC interface via USB port use USB Type A/B cable 22050-60.
Software is supplied in CD-ROM format; compatible with Windows 95/98/NT/2000/XP; Windows XP is recommended for best performance.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Masterflex Linkable Instrument Control Software, CD-ROM
Product Type Software,Brand Masterflex
EW-07551-70
In stock.
$252.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Masterflex Linkable Instrument Control Software (WinLIN)
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Tubing and Pump Accessories
> Masterflex Linkable Instrument Control Software (WinLIN)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity