Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Product Type:
Availability:
In Stock (253)
Control type:
Rotor material:
Housing material:
Max flow rate (LPM):
8.0 (13)
13.0 (8)
42 (6)
2.3 (4)
37 (2)
19.0 (2)
5.3 (1)
35 (1)
17.0 (1)
8 (1)
17 (1)
Material:
IP rating:
IP22 (10)
IP56 (7)
No. of channels:
4 (9)
1 (6)
2 (5)
8 (2)
3 (2)
1 or 2 (1)
1 to 6 (1)
1 to 4 (1)
CE Compliance:
Yes (7)
Max flow rate (GPM):
3.5 (7)
11.1 (6)
2.1 (3)
hp:
1/2 (2)
2,600 pages
of answers

FREE
2013/14 Cole-Parmer
Sourcebook
Over 200 pages of pumping technology including hundreds of pages of flow accessories such as fittings, tubing and process control.
Request your copy today.

Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334

More Unique Lab & Industrial Supplies

 

Masterflex Peristaltic Pumps

Masterflex Peristaltic Pumps

Cole-Parmer offers a range of tubing pumps suitable for dispensing, metering and general transfer applications. Our tubing pumps offer flow rates as low as 0.0007 mL/min to 45 LPM and are able to generate pressures up to 8.6 bar (125 psi).

Tubing pumps confine the media to the tubing providing you with a pump that cannot contaminate the fluid and the fluid cannot contaminate your pump. This contamination free pumping makes tubing pumps great for use in high purity application including the transfer or dosing of chemicals and additives in food, pharmaceutical and semiconductor applications. In addition as the media is confined to the tubing a tubing pump offers easy maintenance and reduced down time compared to other pumping technologies.

Tubing pumps use flexible tubing to run through rollers in the pump head. As fluid goes through the pump head, packets of fluids are formed. Flow rate is determined by the size of the packets and the speed of the rollers turning. By controlling the size of the tubing and the speed of the pump head accurate metering of the media can be achieved.

Cole-Parmer offer tubing pumps suitable for use in labs, pilot plants, process and OEM applications. From low cost fixed speed pumps to advanced controlled models suitable for critical metering and dispensing applications Cole-Parmer is able to offer a tubing pump solution.

Browse through our on-line catalog and use our tools to find your tubing pump or contact our application specialist for free technical support and let us bring over 50 years of pumping experience to solve your application needs.

Advantages for tubing pumps

  • The only part of a tubing pump that comes in contact with the liquid is the tubing. This maintains the sterility of fluid and the pump
  • Cole-Parmer offers over 20 tubing formulations and sizes for their tubing pumps for excellent compatibility with a variety of liquids
  • Self priming - our tubing pump will draw fluid into the tubing when starting dry, up to 8.8 m suction lift. Other pumps require user to fill pump and suction line with fluid before use which can be inconvenient and also create potential for hazardous spills or contamination
  • Gentle pumping action - low shear - makes a tubing pump ideal for handling cells and large proteins, as the pumping action will not damage the content
  • The fluid does not contact the tubing pump so there are no parts to clean such as valves, diaphragms, seals etc. All you have to do is clean or replace the tubing. This easy tubing replacement means reduced maintenance times
  • A tubing pump is non-siphoning which means that they prevent back flow into the system. This promotes accuracy during dispensing


Typical Application for tubing pumps

  • Juice production
  • Pizza sauce dispensing
  • Vitamin A & D Injection
  • Aspiration of tissue culture medium
  • Circulation of cell suspension in fermentation
  • Cosmetic dispensing
  • Harvesting cell media
  • Manufacturing IV bag dispensing
  • Nutrient supply for cultures
  • Sterilized media dispensing
  • Acid/base dispensing
  • Adhesives for cement
  • Circuit board manufacturing
  • Dispensing glue emulsions
  • Transfer of Fuels and lubricants
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
EW-07523-80 Masterflex L/S Digital Drive 07523-80

Masterflex L/S Digital Drive, 600 RPM, 115/230 VAC

Product Type Pump Drive,Max flow rate mL/min 3400,Min flow rate mL/min 0.006
EW-07523-80
In stock.
$1,898.00 USD/EACH

Qty:

Representative photo only Masterflex L S Precision Variable Speed Drive with Remote Input 6 to 600 rpm 90 to 260 VAC

Masterflex L/S Precision Variable-Speed Drive with Remote Input; 6 to 600 rpm, 90 to 260 VAC.

Product Type Pump Drive,Max flow rate mL/min 3400,Min flow rate mL/min 0.36
EW-07528-10
In stock.
$1,120.00 USD/EACH

Qty:

EW-07523-90 Masterflex L/S Digital Drive 07523-90

Masterflex L/S Digital Drive, 100 RPM, 115/230 VAC

Product Type Pump Drive,Max flow rate mL/min 566,Min flow rate mL/min 0.001
EW-07523-90
In stock.
$1,898.00 USD/EACH

Qty:

Representative photo only Masterflex L S Precision Variable Speed Drive with Remote Input 1 to 100 rpm 90 to 260 VAC

Masterflex L/S Precision Variable-Speed Drive with Remote Input; 1 to 100 rpm, 90 to 260 VAC.

Product Type Pump Drive,Max flow rate mL/min 580,Min flow rate mL/min 0.06
EW-07528-30
In stock.
$1,120.00 USD/EACH

Qty:

Representative photo only Masterflex L S Easy Load II pump head for precision tubing adjustable occlusion SS rotor

Masterflex L/S Easy-Load II pump head for precision tubing, adjustable occlusion, SS rotor.

Product Type Pump Head,Rotor material Stainless steel,Housing material Polyphenylene sulfide
EW-77201-60
In stock.
$338.00 USD/EACH

Qty:

Representative photo only Masterflex L S Easy Load II pump head for precision tubing fixed occlusion SS rotor

Masterflex L/S Easy-Load II pump head for precision tubing, fixed occlusion, SS rotor.

Product Type Pump Head,Rotor material Stainless steel,Housing material Polyphenylene sulfide
EW-77200-60
In stock.
$296.00 USD/EACH

Qty:

EW-77410-10 Masterflex<small><sup>&reg;</sup></small> I/P<small><sup>&reg;</sup></small> Precision Brushless Drive, 33 to 650 rpm, 115/230 VAC

Masterflex I/P Precision Brushless Drive, 33 to 650 rpm, 115/230 VAC

Product Type Masterflex I/P Drive,Max flow rate mL/min 19000,Min flow rate mL/min 200
EW-77410-10
In stock.
$2,035.00 USD/EACH

Qty:

Representative photo only Masterflex I P Easy Load pump head PSF housing SS rotor

Masterflex I/P Easy-Load pump head, PSF housing/SS rotor.

Product Type Pump Head,Max flow rate LPM 13.0,Max flow rate GPM 3.5
EW-77601-10
In stock.
$660.00 USD/EACH

Qty:

Representative photo only Masterflex L S economy variable speed drive 20 to 600 rpm 115 VAC

Masterflex L/S economy variable-speed drive, 20 to 600 rpm, 115 VAC.

Product Type Pump Drive,Max flow rate mL/min 2900,Min flow rate mL/min 1.2
EW-07554-90
In stock.
$638.00 USD/EACH

Qty:

EW-77921-70 Masterflex<small><sup>&reg;</sup></small> L/S<small><sup>&reg;</sup></small> Digital Pump System with Easy-Load<small><sup>&reg;</sup></small> II Pump Head, 600 RPM, 115/230 VAC

Masterflex L/S Digital Pump System with Easy-Load II Pump Head, 600 RPM, 115/230 VAC

Product Type Peristaltic Pump,Min flow rate mL/min 0.28,Max flow rate mL/min 1700
EW-77921-70
In stock.
$2,230.00 USD/EACH

Qty:

EW-77601-60 Masterflex<small><sup>&reg;</sup></small> I/P<small><sup>&reg;</sup></small> Easy-Load<small><sup>&reg;</sup></small> pump head,  PPS housing/SS rotor.

Masterflex I/P Easy-Load pump head, PPS housing/SS rotor.

Product Type Pump Head,Max flow rate LPM 13.0,Max flow rate GPM 3.5
EW-77601-60
In stock.
$715.00 USD/EACH

Qty:

Representative photo only Masterflex L S Easy Load II pump head for high performance precision tubing fixed occlusion SS rotor

Masterflex L/S Easy-Load II pump head for high-performance precision tubing, fixed occlusion, SS rotor.

Product Type Pump Head,Rotor material Stainless steel,Housing material Polyphenylene sulfide
EW-77200-62
In stock.
$296.00 USD/EACH

Qty:

Representative photo only L S computer compatible programmable digital drive 0 1 to 600 rpm 115 230 VAC

L/S computer-compatible/programmable digital drive, 0.1 to 600 rpm, 115/230 VAC

Product Type Pump Drive,Max flow rate mL/min 3400,Min flow rate mL/min 0.006
EW-07551-00
In stock.
$2,198.00 USD/EACH

Qty:

EW-07557-00 Masterflex L/S Modular Drive 07557-00

Masterflex L/S Precision Modular Drive with Bench Top Controller and Remote I/O; 6 to 600 rpm, 90 to 260 VAC

Product Type Pump Drive,Max flow rate mL/min 3400,Min flow rate mL/min 0.36
EW-07557-00
In stock.
$1,190.00 USD/EACH

Qty:
See all

Representative photo only Masterflex L S Easy Load pump head for precision tubing PSF housing CRS rotor

Masterflex L/S Easy-Load pump head for precision tubing; PSF housing, CRS rotor.

Product Type Pump Head,Rotor material Cold-rolled steel,Housing material Polysulfone
EW-07518-00
In stock.
$206.00 USD/EACH

Qty:

382 results (showing 1 - 15 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title