Home  >  Clean Room > Mats
Mats
Mats
Government regulations and a need to keep contamination out has us all scrambling for the best ways to cut down on unwarranted particles to preserve our cleanroom environment. Nothing is more important than keeping contaminants out of your cleanroom. That's what makes a cleanroom mat so special. Unlike the welcome mat your family has at the front door, a cleanroom mat is an adhesive mat that grabs dust, dirt, contaminants, and other particles off of your feet. The sticky mats actually capture the dirt and as the cleaning mats get sullied you are able to peel away the dirt exposing a new and fresh clean mat. Some mats include slip-resistant material on the bottom for safety. Other cleanroom mats are used for comfort within the lab and include a grounding snap for static control. A cleanroom mat is a necessary component for any controlled environment.
 Product Detail
Sort
 Availability
 Price
EW-09411-00 Cole-Parmer<small><sup>&reg;</sup></small> adhesive floor mats; 45.5" x 18", 120/box
Cole-Parmer adhesive floor mats; 45.5" x 18", 120/box
Product Type Floor Mats,Description Adhesive mats measure 45.5 in L x 18 in W x.0625 thick.,Material Polyethylene,Color Blue
EW-09411-00
In stock.
$117.00 USD/BOX of 120
Qty:
EW-09411-03 Representative image shown.
Adhesive Floor Mat, 240 Sheet box, 18" x 36"
Product Type Floor Mats,Color Clear on White,Material Polyethylene
EW-09411-03
In stock.
$173.00 USD/CASE
Qty:
Representative photo only Cleanroom Mat 18 x36 30 Layer Pk 8 White
Cleanroom Mat 18"x36", 30 Layer, Pk/8 White
Product Type Floor Mats,Color White,Material LDPE
EW-64100-02
In stock.
$170.00 USD/PACKAGE
Qty:
See all Clean Room Mats
Representative photo only Cleanroom Mat 18 x36 30 Layer Pk 4 White
Cleanroom Mat 18"x36", 30 Layer, Pk/4 White
Product Type Floor Mats,Color White,Material LDPE
EW-64100-01
In stock.
$117.00 USD/PACKAGE
Qty:
See all Clean Room Mats
EW-09411-07 Representative image shown.
Adhesive Floor Mat, 25" x 45", Clear on White, 30-layer mats, 8 mats/case
Product Type Floor Mats,Color Clear on White,Material Polyethylene
EW-09411-07
In stock.
$191.00 USD/CASE
Qty:
Representative photo only Mat Refill 60 Layer Blue Pk 4
Mat Refill 60 Layer Blue Pk 4
Product Type Floor Mats,Description Refill,Material LDPE,Color Blue
EW-64100-47
In stock.
$180.00 USD/PACKAGE
Qty:
Representative photo only Cleanroom Mat 24 x36 30 Layer Pk 4 White
Cleanroom Mat 24"x36", 30 Layer, Pk/4 White
Product Type Floor Mats,Color White,Material LDPE
EW-64100-11
In stock.
$135.00 USD/PACKAGE
Qty:
See all Clean Room Mats
EW-09411-06 Representative image shown.
Adhesive Floor Mat, 36" x 48", Clear on White, 30-layer mats, 8 mats/case
Product Type Floor Mats,Color Clear on White,Material Polyethylene
EW-09411-06
Call 1-888-358-4705 for availability.
$250.00 USD/CASE
Qty:
EW-09411-24 Representative image shown.
Adhesive Floor Mat, 18" x 36", Clear on White, 60-layer mats, 4 mats/case
Product Type Floor Mats,Color White,Material Polyethylene
EW-09411-24
In stock.
$120.00 USD/CASE
Qty:
Representative photo only Mat Refill 60 Layer White Pk 4
Mat Refill 60 Layer White Pk 4
Product Type Floor Mats,Description Refill,Material LDPE,Color White
EW-64100-46
Call 1-888-358-4705 for availability.
$180.00 USD/PACKAGE
Qty:
Representative photo only Adhesive Mat Starter Kit Blue 60 Layer
Adhesive Mat Starter Kit Blue 60 Layer
Product Type Floor Mats,Description Starter Kit,Material LDPE,Color Blue
EW-64100-45
Usually ships in 7 days or sooner.
$117.00 USD/EACH
Qty:
EW-09411-26 Representative image shown.
Adhesive Floor Mat, 36" x 46", Clear on White, 60-layer mats, 4 mats/case
Product Type Floor Mats,Color White,Material Polyethylene
EW-09411-26
In stock.
$237.00 USD/CASE
Qty:
EW-64100-35 Adhesive Mat Starter Kit Blue 30 Layer
Adhesive Mat Starter Kit Blue 30 Layer
Product Type Floor Mats,Description Starter Kit,Material LDPE,Color Blue
EW-64100-35
Usually ships in 12 days.
$91.00 USD/EACH
Qty:
Representative photo only Adhesive Mat Starter Kit White 60 Layer
Adhesive Mat Starter Kit White 60 Layer
Product Type Floor Mats,Description Starter Kit,Material LDPE,Color White
EW-64100-40
In stock.
$117.00 USD/EACH
Qty:
Representative photo only Mat Refill 30 Layer Blue Pk 4
Mat Refill 30 Layer Blue Pk 4
Product Type Floor Mats,Description Refill,Material LDPE,Color Blue
EW-64100-37
Usually ships in 12 days.
$127.00 USD/PACKAGE
Qty:
31 results (showing 1 - 15 )
Per Page 
Home  >  Clean Room > Mats
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title