Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Home  >  Measuring Tools
Measuring Tools
Measuring Tools
Distance meters allow workers to determine precise measurements between two objects. Distance meters come in several different types from a laser distance meter to a sonar or ultrasonic distance meter to even a long range distance meter. Laser distance meters are best when they are accurate to within (+/-) 1 mm and have a range of more than 600 feet. The most popular distance meters sold are the sonar or ultrasonic distance meter. These often have a laser eye used only as a pointing device but use sound waves, not a laser to measure distances. A sonar distance meter is usually precise to within (+/-) 6 inches and up to about 50 feet. There are a variety of brand name distance meters available. At Cole-Parmer you can select from distance meters, NIST-certified straight edge and flexible rules, digital protractors, and steel calibration block kits to meet all of your application requirements. Whether you use standard analog or digital measuring devices, you can rely on the measurements and the simplicity of use helping you reduce errors due to operator interpretation. If you have questions about selecting distance meters for your workspace contact a Cole-Parmer Technical Expert today.
 Product Detail
Sort
 Availability
 Price
52-264012: Fowler 12" /300MM Pre-Certified Flexible Rule
  • Product Type: NIST-Certified Rules
  • Qty/ea: 1
  • Model: 52-264-012
EW-09919-16
In stock.
$67.75 USD/EACH
Qty:
52-411012:Rule 12in/4R Flex Pre-Cert. Ruler Golden Rule Flex
  • Product Type: NIST-Certified Rules
  • Qty/ea: 1
  • Model: 52-411-012
EW-09919-11
In stock.
$67.75 USD/EACH
Qty:
NIST Certified Rigid Rule Set, 6", 12" and 24" Rules.
  • Product Type: NIST-Certified Rules
  • Qty/ea: 1
  • Model: 52-262-624
EW-09919-32
In stock.
$200.00 USD/EACH
Qty:
52-413-012:12in /4R Rigid Pre-Cert. Rule
  • Product Type: NIST-Certified Rules
  • Qty/ea: 1
  • Model: 52-413-012
EW-09919-14
In stock.
$67.75 USD/EACH
Qty:
52-411-006:Rule 6in/4R Flex Pre-Cert. Ruler Golden Rule Flex
  • Product Type: NIST-Certified Rules
  • Qty/ea: 1
  • Model: 52-411-006
EW-09919-00
In stock.
$40.50 USD/EACH
Qty:
Mitutoyo 950-317 Electronic/Digital Protractor
  • Product Type: Protractors
  • Range: 360°
  • Qty/ea: 1
EW-97230-00
In stock.
$329.00 USD/EACH
Qty:
Fowler 53-670-002 Micrometer Calibration Set; 9 Steel Gauge Blocks
  • Product Type: Measuring/Calibrating Kits
  • Qty/ea: 1
  • Model: 53-670-002
EW-97235-02
In stock.
$289.00 USD/EACH
Qty:
52-511-006:6in 5R Flexible Rule Pre-Certified
  • Product Type: NIST-Certified Rules
  • Qty/ea: 1
  • Model: 52-511-006
EW-09919-02
Usually ships in 5 days.
$40.50 USD/EACH
Qty:
52-264-006:6in /150MM Flex Pre-Cert. Rule
  • Product Type: NIST-Certified Rules
  • Qty/ea: 1
  • Model: 52-264-006
EW-09919-06
In stock.
$40.50 USD/EACH
Qty:
Pro-360 Digital Protractor, 360 range
  • Product Type: Protractors
  • Range: 0 to 360 ° (4 x 90 °)
  • Qty/ea: 1
EW-97241-60
In stock.
$291.00 USD/EACH
Qty:
Disto E7100i Handheld Laser Distance meter
  • Product Type: Precison Measurement Tools
  • Range: 0.6 to 200 ft
    (0.18 to 60.9 m)
  • Dimensions: 4 23/32" x 1 15/32" x 29/32"
    (120 x 37 x 23 mm)
EW-97610-33
Usually ships in 4 days.
$161.00 USD/EACH
Qty:
Fowler 54-440-775-1 Digi-Pro Electronic Protractor; 12 inch Blades
  • Product Type: Protractors
  • Range: Full, 360°
  • Power: 3V lithium battery
EW-09917-12
In stock.
$112.00 USD/EACH
Qty:
Fowler Digi-Level Electronic Protractor, 0 to 360 (4 x 90 ) range
  • Product Type: Protractors
  • Range: 0 to 360 ° (4 x 90 °)
  • Description: Fowler Digi-Level Electronic Protractor
EW-09918-71
Usually ships in 4 days.
$162.00 USD/EACH
Qty:
Fowler 52-413-006 NIST-Certified Rule, 6" L Rigid Steel, 4R Intervals
  • Product Type: NIST-Certified Rules
  • Qty/ea: 1
  • Model: 52-413-006
EW-09919-04
Usually ships in 5 days.
$40.50 USD/EACH
Qty:
Fluke-414D Laser Distance Meter; 165 feet (50 meter)
  • Product Type: Distance Measuring Meters
  • Range: 0.05 m to 40 m
    (0.16 ft to 130 ft)
    (for extended distances, use a target plate)
  • Dimensions: 116 mm x 53 mm x 33 mm
    (4.56 in x 2.08 in x 1.29 in)
EW-97241-14
Usually ships in 14 days or sooner.
$139.99 USD/EACH
Qty:
27 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#
#PRODUCTHEADER#
#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Measuring Tools