log in or register
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Safety Products > Metal Detectors
Metal Detectors
Metal Detectors
Metal detectors for food processing are essential. The most common food containment is metal. Metal detectors for food processing ensure that your products, as well as ingredients, are safe and free of any foreign metal contaminants. Food and pharmaceutical processors and use such equipment to help meet stringent industry as well as regulatory compliance (such as HACCP) to make sure products being produced to not pose a hazard to end users. The Thermo Scientific APEX 100 is used for metal detection for food processing and the Thermo Scientific APEX 500 Rx for metal detection in pharmaceutical manufacturing. These units use XR (IXR) technology to accurately detect metal contaminants in wet as well as in dry ingredients and finished products. Installing and setting up the device is quick and simple. All electrical fittings, wiring, and mounting hardware are provided. The intuitive control panel, along with the quick reference guide, allows you to set up and start running samples in no time. When selecting a metal detector consider a couple key factors including type of product for inspection, and current configuration of your conveyor system to help you select the correct opening (or aperture) for the metal detector to help meet your application requirements. If you have questions about metal detectors for food processing contact a Cole-Parmer Technical Expert today.
 Product Detail
Sort
 Availability
 Price
Representative photo only Thermo Scientific APEX 100 Metal Detector with 500 mm width x 200 mm height aperture
Thermo Scientific APEX 100 Metal Detector, with 500 mm width x 200 mm height aperture
Product Type Metal Detectors,Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors,Construction 304 stainless steel straightlined case, ABS plastic control panel
EW-20039-68
Call 1-888-358-4705 for availability.
$10,803.00 USD/EACH
Qty:
Representative photo only Thermo Scientific High Polish Finish for complete system APEX 500 Rx
Thermo Scientific High Polish Finish for complete system APEX 500 Rx
Product Type Accessory,CE Compliance Yes,Brand Thermo Scientific
EW-54502-30
Call 1-888-358-4705 for availability.
$2,500.00 USD/EACH
Qty:
Representative photo only Thermo Scientific APEX 100 Metal Detector with 350 mm width x 200 mm height aperture
Thermo Scientific APEX 100 Metal Detector, with 350 mm width x 200 mm height aperture
Product Type Metal Detectors,Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors,Construction 304 stainless steel straightlined case, ABS plastic control panel
EW-20039-44
Call 1-888-358-4705 for availability.
$10,301.00 USD/EACH
Qty:
Representative photo only Thermo Scientific APEX 100 Metal Detector with 200 mm width x 125 mm height aperture
Thermo Scientific APEX 100 Metal Detector, with 200 mm width x 125 mm height aperture
Product Type Metal Detectors,Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors,Construction 304 stainless steel straightlined case, ABS plastic control panel
EW-20039-17
Call 1-888-358-4705 for availability.
$8,973.00 USD/EACH
Qty:
Representative photo only Thermo Scientific APEX 100 Metal Detector with 600 mm width x 150 mm height aperture
Thermo Scientific APEX 100 Metal Detector, with 600 mm width x 150 mm height aperture
Product Type Metal Detectors,Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors,Construction 304 stainless steel straightlined case, ABS plastic control panel
EW-20039-82
Call 1-888-358-4705 for availability.
$10,579.00 USD/EACH
Qty:
Representative photo only Thermo Scientific APEX 100 Metal Detector with 450 mm width x 200 mm height aperture
Thermo Scientific APEX 100 Metal Detector, with 450 mm width x 200 mm height aperture
Product Type Metal Detectors,Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors,Construction 304 stainless steel straightlined case, ABS plastic control panel
EW-20039-60
Call 1-888-358-4705 for availability.
$10,691.00 USD/EACH
Qty:
Representative photo only Thermo Scientific APEX 100 Metal Detector with 450 mm width x 350 mm height aperture
Thermo Scientific APEX 100 Metal Detector, with 450 mm width x 350 mm height aperture
Product Type Metal Detectors,Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors,Construction 304 stainless steel straightlined case, ABS plastic control panel
EW-20039-63
Call 1-888-358-4705 for availability.
$12,857.00 USD/EACH
Qty:
Representative photo only Thermo Scientific APEX 100 Metal Detector with 550 mm width x 200 mm height aperture
Thermo Scientific APEX 100 Metal Detector, with 550 mm width x 200 mm height aperture
Product Type Metal Detectors,Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors,Construction 304 stainless steel straightlined case, ABS plastic control panel
EW-20039-76
Call 1-888-358-4705 for availability.
$10,996.00 USD/EACH
Qty:
Representative photo only Thermo Scientific APEX 100 Metal Detector with 400 mm width x 200 mm height aperture
Thermo Scientific APEX 100 Metal Detector, with 400 mm width x 200 mm height aperture
Product Type Metal Detectors,Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors,Construction 304 stainless steel straightlined case, ABS plastic control panel
EW-20039-52
Call 1-888-358-4705 for availability.
$10,510.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Rejector with 2 inch 50 8 mm OD for hose attachement for good product flow
Thermo Scientific Rejector with 2 inch (50.8 mm) OD for hose attachement for good product flow
Product Type Accessory,CE Compliance Yes,Brand Thermo Scientific
EW-54502-31
Call 1-888-358-4705 for availability.
$650.00 USD/EACH
Qty:
Representative photo only Thermo Scientific APEX 100 Metal Detector with 600 mm width x 100 mm height aperture
Thermo Scientific APEX 100 Metal Detector, with 600 mm width x 100 mm height aperture
Product Type Metal Detectors,Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors,Construction 304 stainless steel straightlined case, ABS plastic control panel
EW-20039-80
Call 1-888-358-4705 for availability.
$10,116.00 USD/EACH
Qty:
Representative photo only Thermo Scientific APEX 100 Metal Detector with 200 mm width x 350 mm height aperture
Thermo Scientific APEX 100 Metal Detector, with 200 mm width x 350 mm height aperture
Product Type Metal Detectors,Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors,Construction 304 stainless steel straightlined case, ABS plastic control panel
EW-20039-23
Call 1-888-358-4705 for availability.
$10,301.00 USD/EACH
Qty:
Representative photo only Thermo Scientific APEX 100 Metal Detector with 250 mm width x 125 mm height aperture
Thermo Scientific APEX 100 Metal Detector, with 250 mm width x 125 mm height aperture
Product Type Metal Detectors,Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors,Construction 304 stainless steel straightlined case, ABS plastic control panel
EW-20039-25
Call 1-888-358-4705 for availability.
$9,197.00 USD/EACH
Qty:
Representative photo only Thermo Scientific APEX 100 Metal Detector with 550 mm width x 250 mm height aperture
Thermo Scientific APEX 100 Metal Detector, with 550 mm width x 250 mm height aperture
Product Type Metal Detectors,Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors,Construction 304 stainless steel straightlined case, ABS plastic control panel
EW-20039-77
Call 1-888-358-4705 for availability.
$11,737.00 USD/EACH
Qty:
Representative photo only Thermo Scientific APEX 100 Metal Detector with 500 mm width x 150 mm height aperture
Thermo Scientific APEX 100 Metal Detector, with 500 mm width x 150 mm height aperture
Product Type Metal Detectors,Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors,Construction 304 stainless steel straightlined case, ABS plastic control panel
EW-20039-66
Call 1-888-358-4705 for availability.
$10,131.00 USD/EACH
Qty:
123 results (showing 1 - 15 )
Per Page 
Home  >  Safety Products > Metal Detectors
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title