Home  >  Tubing, and Hose > Tubing Cutters > Metallic Tubing Cutter
Metallic Tubing Cutter
Metallic Tubing Cutter
  • Clean, burr-free cuts improve chromatographic reproducibility
  • Cuts metallic tubing faster with more precision
  • Assures zero dead volume tubing connections in chromatography applications
  • Eliminates sample dispersion caused by improper deburring of tubing
Eliminate burrs and uneven cuts on stainless steel and copper tubing for chromatography applications. Clamp vice on swing arm holds tubing securely in place while the abrasive cutting wheel produces a square cut on tubing. Protective shield covers the abrasive wheel for safe operation. Use with any HPLC system or for chromatography pilot scale plumbing.
 Product Detail
Sort
 Availability
 Price
EW-95649-00 Metallic Tubing Cutter, 110-120/220-240 V
Metallic Tubing Cutter, 110-120/220-240 V
Product Type Tubing Cutters
EW-95649-00
In stock.
$960.00 USD/EACH
Qty:
1 results (showing 1 - 1 )
Per Page 
Home  >  Tubing, and Hose > Tubing Cutters > Metallic Tubing Cutter
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title