Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 
Home  >  Methyl Orange-xylene cyanol solution in water

Methyl Orange-xylene cyanol solution in water

Methyl Orange-xylene cyanol solution in water
Choose from our comprehensive line of Methyl Orange-xylene cyanol solution in water and other Methyli to Methylo products.
 Product Detail
Sort
 Availability
 Price

Methyl Orange-xylene cyanol solution in water (500ml)

  • Product Type: Chemical Reagents
  • CAS Number: 62758-15-0
  • Form: Liquid
EW-88351-71
Usually ships in 2 days.
$190.00 USD/EACH

Qty:

1 results (showing 1 - 1 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Methyl Orange-xylene cyanol solution in water