log in or register
Product Type:
Price Range:
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Recorders & Data Acquisition > Dataloggers > Temperature & Humidity Dataloggers > MicroLogPROII Temperature/Humidity/Process Data Loggers
MicroLogPROII Temperature/Humidity/Process Data Loggers
MicroLogPROII Temperature/Humidity/Process Data Loggers
  • Enhanced features including auto e-mail and SMS reporting
  • Compact rugged design for busy areas, laboratories, cold rooms, and more
  • Sample from once per second to once every 18 hours
  • Built-in clock and calendar track your data in real time
  • Data logger kits provide a turn-key solution
The MicroLogPROII data loggers build on the MicroLog products on page 456 by adding enhanced features including modified built-in sensor locations for improved reliability and stabilization time, higher sampling resolution and rates for more accurate readings, USB port for PC communications and firmware updates, and an LED alarm indicator.
Logging capacity is increased to a hefty 52,000 samples of cyclic memory over three channels, allowing you to track important temperature and humidity data for many months. The display is a four-digit LCD with decimal point to show current conditions—minimum and maximum readings are available at the touch of a button. LEDs provide logging and alarm indications if set limits are passed. A unique and powerful new feature is an auto e-mail notification when data is downloaded or when data meets or exceeds user-designed criteria.
Model 35710-93 measures and logs temperature with an internal sensor while model 35710-94 has both temperature and humidity internal sensors. Several external plug-in sensors are available to allow an additional input to either model.
External sensors can be plugged into a port located on the side of each logger. Select from temperature, current, or voltage sensors. Each can be used to log and display additional external readings on your computer.
Temperature sensor is made of stainless steel and can be immersed in liquids or soil or used to measure the temperature of air. Current and voltage sensors can be scaled to match range and units of your transmitter or instrument output. Use the external pH adapter with a standard pH probe (not provided) to log pH, conductivity, and more.
The DataSuite software and cable package (sold separately) lets you create graphs and customize collection parameters including the auto e-mail notification feature.
 Product Detail
Sort
 Availability
 Price
Representative photo only MicroLogPROII Temperature Humidity Data Logger EC850
MicroLogPROII Temperature/Humidity Data Logger EC850
Product Type Temperature/Humidity Dataloggers,Display Four digit, seven segment LCD with decimal point,Interface Mini USB to a PC
EW-35710-94
Call 1-888-358-4705 for availability.
$175.00 USD/EACH
Qty:
Representative photo only MicroLogPROII Temperature Humidity Data Logger EC850 with software and cable
MicroLogPROII Temperature/Humidity Data Logger EC850 with software and cable
Product Type Temperature/Humidity Dataloggers,Display Four digit, seven segment LCD with decimal point,Interface Mini USB to a PC
EW-35710-98
Call 1-888-358-4705 for availability.
$195.00 USD/EACH
Qty:
Representative photo only MicroLogPRO II Temperature Data Logger EC800
MicroLogPRO II Temperature Data Logger EC800
Product Type Temperature Dataloggers,Display Four digit, seven segment LCD with decimal point,Interface Mini USB to a PC
EW-35710-93
Call 1-888-358-4705 for availability.
$135.00 USD/EACH
Qty:
Representative photo only MicroLogPROII Temperature Data Logger EC800 with software and cable
MicroLogPROII Temperature Data Logger EC800 with software and cable
Product Type Temperature Dataloggers,Display Four digit, seven segment LCD with decimal point,Interface Mini USB to a PC
EW-35710-97
Call 1-888-358-4705 for availability.
$155.00 USD/EACH
Qty:
4 results (showing 1 - 4 )
Per Page 
Home  >  Recorders & Data Acquisition > Dataloggers > Temperature & Humidity Dataloggers > MicroLogPROII Temperature/Humidity/Process Data Loggers
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title