log in or register
Brand:
Oakton (1)
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Recorders & Data Acquisition > Dataloggers > Temperature Dataloggers > MicroLog Temperature/Process Data Logger
MicroLog Temperature/Process Data Logger
MicroLog Temperature/Process Data Logger
  • Compact, rugged housing guarantees operation in busy or rugged areas!
  • Automatically log temperature (°F or °C) readings for months, then download data to your computer Sample from once per 10 seconds to once per 2 hours—built-in clock and calendar track your data in real time
This logger stores up to 16,000 measurements, allowing you to track important temperature, and other process data for months. Logger is ideal for shipping, storage, laboratories, factories, refrigerators, freezers, incubators, desiccators, greenhouses, and more. The two-digit LCD shows current conditions—minimum and maximum readings are available at the touch of a button. Display flashes if set limits are passed.
Model 35710-91 records temperature and features a water-resistant and dustproof IP65-rated housing. Optional external sensors (sold separately) allow an additional parameter to be recorded. Connect current and voltage sensors with a transmitter to log other processes such as pressure, flow, pH, and level.
Software lets you set sampling rate, create graphs, and customize logging parameters. Software/cable package is required for use with loggers and are sold separately.
 Product Detail
Sort
 Availability
 Price
Representative photo only MicroLog Temperature Data Logger EC600
MicroLog Temperature Data Logger EC600
Product Type Temperature Dataloggers,Display Two digit LCD display, 0.375 in H,No. of channels 1 internal, 1 external
EW-35710-91
Call 1-888-358-4705 for availability.
$100.00 USD/EACH
Qty:
Representative photo only MicroLog Temperature Data Logger EC600 with software and cable
MicroLog Temperature Data Logger EC600 with software and cable
Product Type Temperature Dataloggers,Display Two digit LCD display, 0.375 in H,No. of channels 1 internal, 1 external
EW-35710-95
In stock.
$120.00 USD/EACH
Qty:
2 results (showing 1 - 2 )
Per Page 
Home  >  Recorders & Data Acquisition > Dataloggers > Temperature Dataloggers > MicroLog Temperature/Process Data Logger
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title